Skip to Content
Merck
All Photos(1)

Key Documents

R3277

Sigma-Aldrich

Rutaecarpine

>98% (HPLC)

Synonym(s):

8,13-Dihydro­indolo[2′,3′:3,4]pyrido[2,1-b]quinazolin-5(7H)-one, Rhetine, Rutecarpine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13N3O
CAS Number:
Molecular Weight:
287.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

>98% (HPLC)

form

solid

color

white

solubility

DMSO: soluble 18 mg/mL (clear yellow solution)
H2O: insoluble

storage temp.

2-8°C

SMILES string

O=C1N2CCc3c([nH]c4ccccc34)C2=Nc5ccccc15

InChI

1S/C18H13N3O/c22-18-13-6-2-4-8-15(13)20-17-16-12(9-10-21(17)18)11-5-1-3-7-14(11)19-16/h1-8,19H,9-10H2

InChI key

ACVGWSKVRYFWRP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Delayed rectifier K+ channel blocker. Inhibits platelet aggregation; vasoldilator.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Máté Bubenyák
Acta pharmaceutica Hungarica, 81(4), 139-149 (2012-02-15)
Quinazolinocarboline rutaecarpine and evodiamine (Evodia rutaecarpa) are main alkaloid components of traditional Chinese folk-remedies. Evodiamine exhibited selective antitumor and antimetastatic effects on several cancer cell lines and became lead structure of anticancer agents. During our synthetic research we achieved to
Shixi Xu et al.
Journal of ethnopharmacology, 139(2), 395-400 (2011-11-29)
Wu-Chu-Yu is a well-known herbal drug used for hypertension. Rutaecarpine and evodiamine are main bioactive components of the medicine. A sensitive and specific HPLC method was developed to analyze rutaecarpine (Rut) and evodiamine (Evo) in rat whole blood. The pharmacokinetics
Chuan-Qin Hu et al.
Journal of Asian natural products research, 14(7), 634-639 (2012-05-16)
A new natural product, 10-hydroxyrutaecarpine (1), and a rarely new glycosidic alkaloid, rutaecarpine-10-O-rutinoside (2), along with the known compounds rutaecarpine (3), evodiamine, wuzhuyuamide-I, and dehydroevodiamine were isolated from the butanol fraction of 70% ethanol aqueous extract of the dried and
Wenguang Yin et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 34(22), 2946-2949 (2010-03-10)
To establish a SPE-HPLC method for the determination and pharmacokinetic study of evodiamine and rutacarpine in rat plasma. A Kromasil C18 column (4.6 mm x 250 mm, 5 microm) was used with acetonitrile-water-tetrahydrofuran-acetic acid (51:48:1:0.1) as a mobile phase and
S N Wu et al.
Neuropharmacology, 41(7), 834-843 (2001-10-31)
The effects of rutaecarpine on ionic currents of NG108-15 neuronal cells were investigated in this study. Rutaecarpine (2-100 microM) suppressed the amplitude of delayed rectifier K+ current (I(K(DR))) in a concentration-dependent manner. The IC50 value for rutaecarpine-induced inhibition of I(K(DR))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service