Skip to Content
Merck
All Photos(1)

Key Documents

R129

Sigma-Aldrich

Monoclonal Anti-Ryanodine Receptor antibody produced in mouse

clone 34C, ascites fluid

Synonym(s):

Anti-CCO, Anti-KDS, Anti-MHS, Anti-MHS1, Anti-PPP1R137, Anti-RYDR, Anti-RYR, Anti-RYR-1, Anti-SKRR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

34C, monoclonal

mol wt

antigen 565 kDa (non-mammalian vertebrates, doublet at 565 kDa representing the α and β isoforms)

species reactivity

human, rat, rabbit, canine, mink, bovine, frog, fish, primate, chicken, mouse, sheep

technique(s)

direct immunofluorescence: suitable
immunohistochemistry (frozen sections): 1:1,000
immunoprecipitation (IP): suitable
western blot: 1:5,000

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The antibody reacts strongly with RyR-1 (expressed predominantly in skeletal muscle and areas of the brain; also known as the α isoform) and RyR-2 (expressed predominantly in the heart muscle, but also found in stomach, endothelial cells and diffuse areas of the brain; also known as the β isoform). In immunohistochemistry, the antibody stains are consistent with the endoplasmic reticulum localization of the receptor in rat hippocampus.
The ryanodine receptor (RyR) is the channel responsible for the release of Ca2+ from the sarcoplasmic reticulum (SR) in muscle cells and it also plays a role in Ca2+ regulation in non-muscle cells. The RyR exists as a homotetramer and is predicted to have a short cytoplasmic C-terminus and 4-10 transmembrane domains; the remainder of the protein, termed the "foot" region is located in the cytoplasm between the T-tubule and the SR. The mammalian RyR is the product of three different genes: RyR-1, which is expressed predominantly in skeletal muscle and areas of the brain, RyR-2, which is expressed predominantly in the heart muscle but also found in the stomach, endothelial cells and diffuse areas of the brain, and RyR-3 which is found in smooth muscle and the brain (striatum, thalamus and hippocampus).

Immunogen

partially purified chicken pectoral muscle ryanodine receptors (RyR)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Mouse monoclonal clone 34C anti-Ryanodine receptor is used to tag ryanodine receptors for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of ryanodine receptor in studies of calcium flux from the sarcoplasmic reticulum of muscle cells.

Physical form

Purified IgG1 antibody containing 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele Scorzeto et al.
PloS one, 8(10), e74919-e74919 (2013-10-08)
Mitochondrial calcium handling and its relation with calcium released from sarcoplasmic reticulum (SR) in muscle tissue are subject of lively debate. In this study we aimed to clarify how the SR determines mitochondrial calcium handling using dCASQ-null mice which lack
Masahiro Aoyama et al.
Journal of cell science, 117(Pt 13), 2813-2825 (2004-06-01)
Intracellular Ca(2+) ([Ca(2+)](i)) oscillations seen in interstitial cells of Cajal (ICCs) are considered to be the primary pacemaker activity in the gut. Here, we show evidence that periodic Ca(2+) release from intracellular Ca(2+) stores produces [Ca(2+)](i) oscillations in ICCs, using
Janet E Jeffrey et al.
Arthritis research & therapy, 9(6), R129-R129 (2007-12-22)
This study investigated the release of prostaglandin E2 (PGE2) from cartilage following an impact load in vitro and the possible chondroprotective effect of cyclooxygenase-2 (COX-2) inhibition using non-steroidal anti-inflammatory drugs (NSAIDs). Explants of human articular cartilage were subjected to a
Katja Anttila et al.
Acta histochemica et cytochemica, 40(2), 35-41 (2007-06-20)
In this study, the fiber type specificity of dihydropyridine receptors (DHPRs) and ryanodine receptors (RyRs) in different rat limb muscles was investigated. Western blot and histochemical analyses provided for the first time evidence that the expression of both receptors correlates
Ken'ichiro Nogami et al.
BMC musculoskeletal disorders, 21(1), 479-479 (2020-07-23)
Previous research indicated that nitric oxide synthase (NOS) is the key molecule for S-nitrosylation of ryanodine receptor 1 (RyR1) in DMD model mice (mdx mice) and that both neuronal NOS (nNOS) and inducible NOS (iNOS) might contribute to the reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service