Skip to Content
Merck
All Photos(3)

Key Documents

R0533

Sigma-Aldrich

Rifapentine

Synonym(s):

3-(((4-cyclopentyl-1-piperazinyl)imino)methyl)rifamycin, Cyclopentylrifampicin, DL 473

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H64N4O12
CAS Number:
Molecular Weight:
877.03
EC Number:
UNSPSC Code:
51283603
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

Assay

≥93.5%

form

powder or crystals

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria

Mode of action

enzyme | inhibits

shipped in

wet ice

storage temp.

2-8°C

SMILES string

N5(CCN(CC5)C6CCCC6)N=Cc1c2c(c3c(c4c(c(c3O)C)O[C@](O\C=C\C([C@H]([C@H]([C@@H]([C@@H]([C@@H]([C@H]([C@H](\C=C\C=C(/C(=O)N2)\C)C)O)C)O)C)OC(=O)C)C)OC)(C4=O)C)c1O)O

InChI

1S/C47H64N4O12/c1-24-13-12-14-25(2)46(59)49-37-32(23-48-51-20-18-50(19-21-51)31-15-10-11-16-31)41(56)34-35(42(37)57)40(55)29(6)44-36(34)45(58)47(8,63-44)61-22-17-33(60-9)26(3)43(62-30(7)52)28(5)39(54)27(4)38(24)53/h12-14,17,22-24,26-28,31,33,38-39,43,53-57H,10-11,15-16,18-21H2,1-9H3,(H,49,59)/b13-12+,22-17+,25-14-,48-23?/t24-,26+,27+,28+,33?,38-,39+,43+,47-/m0/s1

InChI key

WDZCUPBHRAEYDL-OABFQHKQSA-N

General description

Chemical structure: macrolide

Application

Rifapentine is an antibiotic clinically used to treat tuberculosis. It is used in tuberculosis research[1].

Biochem/physiol Actions

Rifapentine is a semisynthetic derivative of rifampicin with antibacterial activity against Gram-positive and Gram-negative bacteria and against Mycobacterium tuberculosis. Rifapentine inhibits DNA-dependant RNA polymerase and prevents RNA transcription. It interacts with bacterial RNA polymerase but does not inhibit the mammalian enzyme[1].
Semisynthetic derivative of rifampicin with antibacterial activity against Gram-positive and Gram-negative bacterial and against Mycobacterium tuberculosis. Rifapentine inhibits DNA-dependant RNA polymerase and prevents RNA transcription.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruth N Moro et al.
Annals of the American Thoracic Society, 15(5), 570-580 (2018-02-03)
Data are limited regarding the safety of 12-dose once-weekly isoniazid (H, 900 mg) plus rifapentine (P, 900 mg) (3HP) for latent infection treatment during pregnancy. To assess safety and pregnancy outcomes among pregnant women who were inadvertently exposed to study
Kathy Williams et al.
Antimicrobial agents and chemotherapy, 56(6), 3114-3120 (2012-04-04)
Novel oral regimens composed of new drugs with potent activity against Mycobacterium tuberculosis and no cross-resistance with existing agents are needed to shorten and simplify treatment for both drug-susceptible and drug-resistant tuberculosis. As part of a continuing effort to evaluate
Simbarashe P Zvada et al.
Antimicrobial agents and chemotherapy, 56(8), 4471-4473 (2012-05-16)
We described the population pharmacokinetics of moxifloxacin and the effect of high-dose intermittent rifapentine in patients with pulmonary tuberculosis who were randomized to a continuation-phase regimen of 400 mg moxifloxacin and 900 mg rifapentine twice weekly or 400 mg moxifloxacin
Kelly E Dooley et al.
The lancet. HIV, 7(6), e401-e409 (2020-04-03)
Short-course preventive therapy with 12 doses of once-weekly rifapentine (900 mg) plus isoniazid (900 mg) could greatly improve tuberculosis control, especially in areas with high co-endemicity with HIV. However, a small previous trial of such therapy with dolutegravir in healthy
Susan E Dorman et al.
The Journal of infectious diseases, 206(7), 1030-1040 (2012-08-02)
Rifapentine administered 5 days per week has potent activity in mouse models of antituberculosis chemotherapy, but efficacy and safety data are limited in humans. We compared the antimicrobial activity and safety of rifapentine vs rifampin during the first 8 weeks

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service