Skip to Content
Merck
All Photos(1)

Key Documents

R0382

Sigma-Aldrich

Retrorsine

≥90% (HPLC), powder, retronecine-type pyrrolizidine alkaloid

Synonym(s):

Retrorsin, Senecionan-11,16-Dione, 12,18-Dihydroxy- (9CI), β-Longilobine, 12,18-Dihydroxysenecionan-11,16-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO6
CAS Number:
Molecular Weight:
351.39
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Retrorsine, ≥90% (HPLC)

Assay

≥90% (HPLC)

mp

208-211 °C (lit.)

SMILES string

C\C=C1\C[C@@H](C)[C@](O)(CO)C(=O)OCC2=CCN3CC[C@@H](OC1=O)[C@@H]23

InChI

1S/C18H25NO6/c1-3-12-8-11(2)18(23,10-20)17(22)24-9-13-4-6-19-7-5-14(15(13)19)25-16(12)21/h3-4,11,14-15,20,23H,5-10H2,1-2H3/b12-3-/t11-,14-,15-,18-/m1/s1

InChI key

BCJMNZRQJAVDLD-CQRYIUNCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Retrorsine has been used:
  • as a mito-inhibitory pyrrolizidine alkaloid compound to induce necrotic liver injury in rats
  • to induce hepatocellular injury in rats
  • to arrest endogenous hepatocyte growth in mice

Biochem/physiol Actions

Retrorsine (RTS) is a retronecine-type pyrrolizidine alkaloid associated with Senecio and Crotalaria species. It belongs to the pyrrolizidine alkaloid family with mito-inhibitory property and elicits hepatotoxicity. It mediates the inactivation of cytochrome P450 3A4. Retrorsine also inhibits replication of fully differentiated hepatocytes.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valerie Gouon-Evans et al.
Nature biotechnology, 24(11), 1402-1411 (2006-11-07)
When differentiated in the presence of activin A in serum-free conditions, mouse embryonic stem cells efficiently generate an endoderm progenitor population defined by the coexpression of either Brachyury, Foxa2 and c-Kit, or c-Kit and Cxcr4. Specification of these progenitors with
Suchitra Sumitran-Holgersson et al.
Cell transplantation, 18(2), 183-193 (2009-06-09)
Although the appearance of hepatic foci in the pancreas has been described in animal experiments and in human pathology, evidence for the conversion of human pancreatic cells to liver cells is still lacking. We therefore investigated the developmental plasticity between
Chun-Hsien Yu et al.
Cell transplantation, 18(10), 1081-1092 (2009-08-05)
Efficient repopulation by transplanted hepatocytes in the severely injured liver is essential for their clinical application in the treatment of acute hepatic failure. We studied here whether and how the transplanted hepatocytes are able to efficiently repopulate the toxin-induced acute
Mabel Fragoso-Serrano et al.
Journal of natural products, 75(5), 890-895 (2012-05-04)
Commercial preparations of the Mexican herbal drug known as "miracle tea" (Packera candidissima and P. bellidifolia) have been profiled qualitatively by HPLC and GC-MS. Eremophilanes (3-7) were the major components found in the hexane-soluble fraction, while pyrrolizidine alkaloids (PAs) were
Ya-Hui Chen et al.
Hepatology (Baltimore, Md.), 57(3), 1215-1224 (2012-10-20)
The potential lineage relationship between hepatic oval cells, small hepatocyte-like progenitor cells (SHPCs), and hepatocytes in liver regeneration is debated. To test whether mature hepatocytes can give rise to SHPCs, rats with dipeptidyl peptidase IV (DPPIV) chimeric livers, which harbored

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service