Skip to Content
Merck

R0141

Floating microcentrifuge tube rack

white

Synonym(s):

tuba storage, tube rack, Nalgene 5974-0404, tube holder, tube organiser

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
41122804
Manufacturer/tradename:
Nalgene® 5974-0404
Feature:
Holds 16 x 1-2 mL tubes
Material:
polypropylene
Packaging:
pack of 4 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene

description

Autoclavable

feature

Holds 16 x 1-2 mL tubes

packaging

pack of 4 ea

manufacturer/tradename

Nalgene® 5974-0404

size

103 mm × 103 mm × 65 mm

color

white

General description

On benchtop, legs prevent tubes from touching surface. In water bath, racks will float with a full load of tubes.

Features and Benefits

  • Floating microcentrifuge tube racks are made of high-quality polypropylene.
  • Square-styled racks are ideal for use in genetic cloning, microbiology, and immunology labs.
  • Floating racks maintain stability and facilitates floatation with a full load of tubes in turbulent water baths.
  • Each hole has molded-in letters and numbers for sample identification.Features a sturdy handle for convenient transport, agitation, and removal from the water bath.
  • On benchtop, legs prevent tubes from touching the surface.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library