Skip to Content
Merck
All Photos(1)

Key Documents

Q1126

Sigma-Aldrich

Q Sepharose Fast Flow

preswollen, 45-165 μm (wet), exclusion limit ~4,000,000 Da

Synonym(s):

Q Sepharose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

form

preswollen

Quality Level

technique(s)

RNA extraction: suitable

particle size

45-165 μm (wet)

pore size

~4,000,000 Da exclusion limit

capacity

0.18-0.24 mmol Cl-/mL, packed gel

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

−Ο−CH2CHOHCH2OCH2CHOHCH2N+(CH3)3

Application

Q Sepharose is used in protein chromatography, ion exchange chromatography and anion exchange media. Q Sepharose has been used to study compounds of plant defense with applications for natural control of phytopathogenic fungi. Q Sepharosehas also been used to develop an efficient method for extracting high-quality mRNA from soil and to study the immunomodulatory proteins from garlic (Allium sativum).

Physical form

Preswollen in 20% ethanol

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 - 109.4 °F - closed cup

Flash Point(C)

38 - 43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E G Campos et al.
International journal for parasitology, 29(5), 655-662 (1999-07-15)
Cytochrome c peroxidase oxidises hydrogen peroxide using cytochrome c as the electron donor. This enzyme is found in yeast and bacteria and has been also described in the trematodes Fasciola hepatica and Schistosoma mansoni. Using partially purified cytochrome c peroxidase
Shikimate dehydrogenase from pepper (<I>Capsicum annuum</I>) seedlings. Purification and properties.
Diaz, J., and Merino, F.
Physiologia Plantarum, 100(1), 147-152 (1997)
Fatima Clement et al.
International immunopharmacology, 10(3), 316-324 (2009-12-17)
Garlic (Allium sativum), an important medicinal spice, displays a plethora of biological effects including immunomodulation. Although some immunomodulatory proteins from garlic have been described, their identities are still unknown. The present study was envisaged to isolate immunomodulatory proteins from raw
M Meilinger et al.
The Journal of biological chemistry, 274(53), 38091-38096 (1999-12-23)
Complement component 3 (C3) and alpha(2)-macroglobulin evolved from a common, evolutionarily old, ancestor gene. Low density lipoprotein-receptor-related protein/alpha(2)-macroglobulin receptor (LRP/alpha(2)MR), a member of the low density lipoprotein receptor family, is responsible for the clearance of alpha(2)-macroglobulin-protease complexes. In this study
B Oster et al.
The Journal of general virology, 89(Pt 1), 87-96 (2007-12-20)
Here, we demonstrate that human herpesvirus 6B (HHV-6B) infection upregulates the tumour suppressor p53 and induces phosphorylation of p53 at Ser392. Interestingly, phosphorylation at the equivalent site has previously been shown to correlate with p53 tumour suppression in murine models.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service