Skip to Content
Merck
All Photos(1)

Key Documents

PZ0298

Sigma-Aldrich

PF-03084014 hydrobromide

≥98% (HPLC)

Synonym(s):

(2S)-2-[[(2S)-6,8-Difluoro-1,2,3,4-tetrahydro-2-naphthalenyl]amino]-N-[1-[2-[(2,2-dimethylpropyl)amino]-1,1-dimethylethyl]-1H-imidazol-4-yl]-pentanamide hydrobromide, Nirogacestat, PF 03084014 hydrobromide, PF 3084014 hydrobromide, PF-3084014 hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H41F2N5O · HBr
CAS Number:
Molecular Weight:
570.56
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 1 mg/mL, clear

storage temp.

room temp

SMILES string

FC1=C2C(CC[C@H](N[C@@H](CCC)C(NC3=CN(C(C)(C)CNCC(C)(C)C)C=N3)=O)C2)=CC(F)=C1.C

InChI

1S/C27H41F2N5O.CH4/c1-7-8-23(32-20-10-9-18-11-19(28)12-22(29)21(18)13-20)25(35)33-24-14-34(17-31-24)27(5,6)16-30-15-26(2,3)4;/h11-12,14,17,20,23,30,32H,7-10,13,15-16H2,1-6H3,(H,33,35);1H4/t20-,23-;/m0./s1

InChI key

GLEYBLZDOTUZJH-WCRWPNQISA-N

Biochem/physiol Actions

PF-03084014 inhibits the Notch signalling pathway. It contributes totumor suppression in breast, pancreatic carcinoma, hepatocellular carcinoma and progressive desmoid tumors. PF-03084014 in combination with dexamethasone elicits antileukemic effects in T-cell acute lymphoblastic leukemias (T-ALL).
PF-03084014 is a highly potent γ-secretase inhibitor that reduces Aβ production in vitro. PF-03084014 does not show a paradoxical increase in plasma Aβ at low concentrations in both preclinical species and humans.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preclinical analysis of the gamma-secretase inhibitor PF-03084014 in combination with glucocorticoids in T-cell acute lymphoblastic leukemia
Samon JB, et al.
Molecular Cancer Therapeutics, 11(7), 1565-1565 (2012)
Clinical activity of the gamma-secretase inhibitor PF-03084014 in adults with desmoid tumors (aggressive fibromatosis)
Kummar S, et al.
Journal of Clinical Oncology, 35(14), 1561-1561 (2017)
Notch inhibitor PF-03084014 inhibits hepatocellular carcinoma growth and metastasis via suppression of cancer stemness due to reduced activation of Notch1-Stat3
Wu CX, et al.
Molecular Cancer Therapeutics, 16(8), 1531-1543 (2017)
Notch pathway inhibition using PF-03084014, a gamma-secretase inhibitor (GSI), enhances the antitumor effect of docetaxel in prostate cancer
Cui D, et al.
Clinical Cancer Research, 21(20), 4619-4629 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service