Skip to Content
Merck
All Photos(3)

Key Documents

PRS2283

Sigma-Aldrich

Anti-RIP3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Receptor-Interacting serine-threonine kinase 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

predicted mol wt 57 kDa

species reactivity

rat, human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ripk3(56532)

Related Categories

Immunogen

a peptide corresponding to amino acids 473-486 of murine RIP3.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)

Linkage

The action of this antibody can be blocked using blocking peptide SBP2283.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xia-Lian Wu et al.
Nature communications, 12(1), 1627-1627 (2021-03-14)
RIPK3 amyloid complex plays crucial roles during TNF-induced necroptosis and in response to immune defense in both human and mouse. Here, we have structurally characterized mouse RIPK3 homogeneous self-assembly using solid-state NMR, revealing a well-ordered N-shaped amyloid core structure featured
Imene Jaadane et al.
Scientific reports, 10(1), 6733-6733 (2020-04-23)
Exposure Limit Values (ELV) for artificial lighting were defined in order to prevent light-induced damage to the retina. The evaluation of the lighting devices include the correction of their spectra by the B(λ) function or blue light hazard function, representing
Douglas Ruhl et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(15), 2951-2964 (2019-02-09)
Ototoxic side effects of cisplatin and aminoglycosides have been extensively studied, but no therapy is available to date. Sensory hair cells, upon exposure to cisplatin or aminoglycosides, undergo apoptotic and necrotic cell death. Blocking these cell death pathways has therapeutic
Alejandra Daruich et al.
Science advances, 5(1), eaau9940-eaau9940 (2019-01-22)
In retinal detachment (RD), photoreceptor death and permanent vision loss are caused by neurosensory retina separating from the retinal pigment epithelium because of subretinal fluid (SRF), and successful surgical reattachment is not predictive of total visual recovery. As retinal iron
Lvshuang Liao et al.
Acta biochimica et biophysica Sinica, 49(10), 879-889 (2017-10-06)
Receptor-interacting protein 3 (RIP3) is an essential component of the necroptosis signaling pathway. Phosphorylation of its downstream target, mixed lineage kinase domain-like protein (MLKL), has been proposed to induce necroptosis by initiating Ca2+ influx. Our previous studies have shown that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service