Skip to Content
Merck

Skip To

PP0540

GlycoProfile β-Elimination Kit

Sufficient for 24 samples

Synonym(s):

β-Elimination Glycan Kit, Glycan Removal Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.28
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

(O-linked)

Quality Segment

storage temp.

room temp

Application

The GlycoProfile β-Elimination kit contains a novel non-reducing reagent to efficiently and specifically remove O-linked carbohydrates from glycoproteins with minimal protein or glycan destruction. Consequently, additional downstream proteomics and glycomic analyses may be employed.

Preparation Note

The kit includes the reagents and techware for O-deglycosylation of up to 24 samples (200 uL at 1-10 ug/uL of glycoprotein per sample).

Legal Information

GlycoProfile is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MDT100MAK175MAK226
Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

-

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

conjugate

(O-linked)

conjugate

-

conjugate

-

conjugate

-


Kit Components Only

Product No.
Description

  • Sodium hydroxide solution, 5.0 M 60 μL

  • Microcon centrifugal filter unit, YM-10 membrane, NMWCO 10 kDa 24 ea

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Centrifugal Filter Unit
    SDS

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

flash_point_f

82.0 °F - closed cup

flash_point_c

27.79 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters