Skip to Content
Merck
All Photos(2)

Key Documents

PLA0238

Sigma-Aldrich

Rabbit anti-AKT1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

AKT, AKT1m, CWS6, PKB, PKB alpha, PKB-ALPHA, PRKBA, Protein kinase B, RAC, RAC-ALPHA, RAC-PK-alpha, protein kinase B alpha, proto-oncogene c-Akt, rac protein kinase alpha, serine-threonine protein kinase, v-akt murine thymoma viral oncogene homolog 1, v-akt murine thymoma viral oncogene-like protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:2,000
western blot: 1:2,000- 1:10,000

accession no.

NP_005154.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... AKT1(207)

Immunogen

The epitope recognized by PLA0238 maps to a region between residue 25 and 75 of human v-akt murine thymoma viral oncogene homolog 1 using the numbering given in entry NP_005154.1 (GeneID 207).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

AKT1 is a serine-threonine kinase that functions downstream of PI-3 kinase and is known to phosphorylate several proteins. AKT1 mediates the effects of several growth factors such as insulin, IGF-1, EGF, and PDGF to control cell survival and apoptosis. There are numerous substrates of AKT1, some of which are: GSK3-alpha and beta, mTOR, TSC2, Bad, c-Raf, and caspase 9.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service