Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
1
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse, human
concentration
1 mg/mL
technique(s)
immunohistochemistry: 1:1,000-1:5,000, immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:2,000-1:10,000
accession no.
NP_002408.2
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... MKI67(4288)
Immunogen
Antibody was affinity purified using an epitope specific to Ki-67 immobilized on solid support. The epitope recognized by PLA0228 maps to a region between residues 1800 and 1850 of human Ki-67 using the numbering given in entry NP_002408.2 (GeneID 4288).
Physical form
Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide
Other Notes
Ki-67 is a marker of cell proliferation detected in cells that are in G1-, S-, G2-, and M-phase of the cell cycle and absent in cells in G0. Ki-67 is detected exclusively in the nucleus, and its detection by immunohistological methods has proven to have diagnostic and prognostic value in the study of a variety of human tumors. The specific cellular functions of Ki-67 have not been elucidated, but it is proposed to function in the organization and maintenance of the architecture of DNA and in the synthesis of ribosomes during mitosis.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.