Skip to Content
Merck

PLA0179

Rabbit anti-NBS1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

AT-V1, AT-V2, ATV, NBS, NBS1, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, P95, cell cycle regulatory protein p95, p95 protein of the MRE11/RAD50 complex

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: 1:100- 1:500, immunoprecipitation (IP): 2- 5 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_002476.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... NBS1(4683)

Immunogen

The epitope recognized by PLA0179 maps to a region between residue 704 and 754 of human Nibrin using the numbering given in entry NP_002476.2 (GeneID 4683).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Mutations in NBS1 (Nijmegen Breakage Syndrome 1), also known as nibrin, are associated with the autosomal recessive syndrome, Nijmegen breakage syndrome, characterized by microcephaly, growth retardation, immunodeficiency, and cancer predisposition. At the cellular level, NBS1 is part of the MRE11/RAD50 double-strand break repair complex that is critical to the DNA damage response, DNA recombination, telomere integrity, and cell cycle checkpoint control.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library