Skip to Content
Merck

Skip To

PKH26GL

PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling

Distributed for Phanos Technologies

Synonym(s):

Red PKH membrane dye

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 kit

Quality Segment

manufacturer/tradename

Distributed for Phanos Technologies

storage condition

protect from light

fluorescence

λex 551 nm; λem 567 nm (PKH26 dye)

detection method

fluorometric

shipped in

ambient

storage temp.

room temp

General description

PKH26 is a red‐fluorescent dye. This lipophilic membrane dye is widely used to determine cell proliferation in heterogeneous cells by flow cytometry.

Application

This kit is for general cell membrane labeling. It has been characterized in a number of model systems and has been found to be useful for in vitro cell labeling, in vitro proliferation studies and long term, in vivo cell tracking. The half-life for elution of PKH26 from labeled rabbit red blood cells is greater than 100 days. This enhanced stability is favorable for long term in vivo studies.
PKH26 Red Fluorescent Cell Linker Kit is for general cell membrane labeling. It has been characterized in a number of model systems and has been found to be useful for in vitro cell labeling, in vitro proliferation studies and long term, in vivo cell tracking.[1] The half-life for elution of PKH26 from labeled rabbit red blood cells is greater than 100 days. This enhanced stability is favorable for long term in vivo studies.

Other Notes

For additional technical details on PKH and CellVue® Fluorescent Cell Linker Dyes including an extensive bibliography, please visit here.

Legal Information

CellVue is a registered trademark of Phanos Technologies

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PKH26PCLMINI26MINI67
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

packaging

pkg of 1 kit

packaging

-

packaging

pkg of 1 kit

packaging

pkg of 1 kit

shipped in

ambient

shipped in

-

shipped in

ambient

shipped in

ambient

storage condition

protect from light

storage condition

-

storage condition

protect from light

storage condition

protect from light

manufacturer/tradename

Distributed for Phanos Technologies

manufacturer/tradename

-

manufacturer/tradename

Distributed for Phanos Technologies

manufacturer/tradename

Distributed for Phanos Technologies

storage temp.

room temp

storage temp.

-

storage temp.

room temp

storage temp.

room temp


Kit Components Only

Product No.
Description

  • Diluent C 6 x 10

  • PKH26 Cell Linker in ethanol .5 mL

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

57.2 °F - closed cup

flash_point_c

14.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 11 Questions  
  1. Can we stain extracellular vesicles particularly exosomes (isolated and suspended in PBS) we have used exosome isolation kit , is ultracentrifugation step must at the end, as for now we are not using ultracentifuge

    1 answer
    1. PKH dyes have been successfully used to label exosomes and extracellular vesicles for in vitro and in vivo tracking experiments. The exosome labeling protocol below details steps for reliable fluorescent labeling of exosomes using PKH dyes for micro vesicle tracking experiments. Instead of PBS, use the kit’s recommended Diluent C for optimal dye performance. Final resuspension in PBS is acceptable for downstream applications.

      Please check this link for more details:
      https://www.sigmaaldrich.com/technical-documents/protocols/biology/cell-culture/exosome-labeling-pkh.html

      These links could also be helpful to you:
      https://www.sigmaaldrich.com/catalog/product/sigma/mini26?lang=en&region=US
      https://www.sigmaaldrich.com/technical-documents/articles/biowire/cell-tracking-lipophilic-membrane-dyes.html

      Helpful?

  2. If we want to use the PKH to label the membrane of fibroblast, and then co-culture them with tumor cells in order to interact, ¿ Will PKH transfer from fibroblast to tumor cells?

    1 answer
    1. PKH dyes are routinely used in co-culture situations. Because staining is nearly instantaneous, rapid and homogeneous dispersion of cells in dye solution is essential. There should be no excess or unincorporated dye in the medium prior to mixing or co-culturing the cell lines. Free dye will be taken up by all cells. See the links below to the product protocol as well as two additional co-culture studies using PKH dyes:

      Protocol
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/271/475/mini26bul.pdf

      Co-culture in scaffolds
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC5155425/

      Co-culture, mixed monolayer, and on Transwell
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC5168417/

      Helpful?

  3. Are cells lost during the PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling staining process?

    1 answer
    1. Over-labeling of the cells will result in loss of membrane integrity and reduced cell recovery.  Methods for improving cell viability can be found on the troubleshooting guide.

      Helpful?

  4. Product PKH26GL, PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, is a red dye.  Do you offer a kit that stains the cells green?

    1 answer
    1. We offer Product numbers:PKH2GL Both of these kits stain the cells with a dye that emits in the green fluorescent range.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. When using Product PKH67GL, PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling, how long will the cells remain stained after treatment?

    1 answer
    1. Labeled cells that have been washed can be visualized in culture up to 100 days after staining (for non-dividing cells). The dye itself is stable and will divide equally when the cells divide. After staining with PKH dyes, you can observe as many as 8 divisions depending on how brightly the cells were stained initially and the amount of surface area on the cells. Most commonly, 4-6 divisions can be visualized.

      Helpful?

  7. Does the dye leak from the cells after labeling when using Product PKH67GL, PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling?

    1 answer
    1. In general, slow loss of fluorescence has been observed with PKH1 and PKH2 in in vivo studies and it is likely that PKH67 may also slowly lose fluorescence. Leakage (or loss of staining intensity) may also occur due to cell-to-cell transfer of the dye. The most common cause of cell-to-cell dye transfer is inadequate washing of the cells. Wash cells 3-5 times after labeling and transfer samples to new tubes between washes.

      Helpful?

  8. What is the difference between PKH26 Red Fluorescent Cell Linker Kit for Phagocytic Cell Labeling Products PKH26GL and PKH26PCL?

    1 answer
    1. The general labeling kit (Product No. PKH26GL) will label the membrane of the cells.  For phagocytic cells, Product No. PKH26PCL is used.  This kit utilizes dye aggregates for specific labeling of phagocytic cells in the presence of other cell types.

      Helpful?

  9. How many cells can be stained with PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Products PKH26GL and MINI26?

    1 answer
    1. For Product PKH26GL, when using the concentrations indicated in the product bulletin, you can stain 30 samples (1x107 cells/sample).The kit provides 500 ul of PKH26 dye solution.  You use 4 ul per sample.  There is enough dye in the kit to stain 125 samples.The kit provides 60 mls of Diluent C.  You use 2 mls per sample (1 ml for dilution of the dye, 1 ml for dilution of the cells).  There is enough diluent for 30 samples.You can purchase additional diluent as Product CGLDILIf you need less dye, you can purchase Product MINI26 which contains 100 ul of dye and 10 mls of Diluent C.  This kit contains enough dye to stain 25 samples and enough diluent to stain 5 samples.

      Helpful?

  10. What method of fixation can be used for tissue/cells with the PKH Fluorescent Cell Linker Kit for General Cell Membrane Labeling dyes?

    1 answer
    1. A protocol for visualization of tissue sections can be found in the technical bulletin.  For visualization of stained cells by immunofluorescence or flow cytometry, the cells can be fixed in 2% paraformaldehyde for 15 minutes. The use of other organic solvents will extract the dye from the cells.  If internal labeling is desired, the cells can be permeabilized with saponin (50-75 μg/mL). Here is the link to the Troubleshooting Guide.

      Helpful?

1–10 of 11 Questions  

Reviews

No rating value

Active Filters