Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.51
UNSPSC Code:
12352200
Form:
solution
Application
PEXBUFF transfection enhancer increases the efficiency of CRISPR based RNP gene editing experiments. When added during RNP preparation workflow, PEXBUF increases editing.
Features and Benefits
- GRAS: generally regarded as safe
- Increases gene editing rates
- Easy workflow addition
- Works with various CAS9-based editors
Disclaimer
This product is covered by PCT/US2022/079037 and sold for research use only. Commercial use will require a license from Sigma-Aldrich. For licensing questions, please contact [email protected].
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| PEXBUF-125UL | 04065270963253 |
| PEXBUF-25UL | 04065270963260 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service