Skip to Content
Merck
All Photos(1)

Documents

P9155

Sigma-Aldrich

Poly-ʟ-Lysine Hydrobromide

synthetic, mol wt 30,000-70,000, powder, γ-irradiated, suitable for cell culture, BioXtra

Synonym(s):

PDL HBr

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.75

product name

Poly-L-lysine hydrobromide, mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture

biological source

synthetic (organic)

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

mol wt

30,000-70,000

packaging

pkg of 5 mg

technique(s)

cell culture | mammalian: suitable

surface coverage

4 μg/cm2

solubility

H2O: soluble 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

Cl.NCCCCC(N)C(O)=O

InChI

1S/C18H38N6O4/c19-10-4-1-7-13(22)16(25)23-14(8-2-5-11-20)17(26)24-15(18(27)28)9-3-6-12-21/h13-15H,1-12,19-22H2,(H,23,25)(H,24,26)(H,27,28)/t13-,14-,15-/m0/s1

InChI key

WBSCNDJQPKSPII-KKUMJFAQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties. Poly-L-lysine hydrobromide is a nonspecific attachment factor in promoting cell adhesion to the substratum.

Application

Recommended as a cell culture substratum when using 0.5 ml of a 0.1 mg/ml solution to coat 25 cm2. Optimal conditions for attachment must be determined for each cell line and application.
Poly-L-lysine hydrobromide has been used to adhere the dissociated cells of cortical tissues into the culture plates. It has also been used to treat slides, to fix samples on it for immunohistochemical analysis.

Biochem/physiol Actions

Poly-L-lysine is employed as a coating substance to facilitate improved cell attachment to culture surfaces. Research indicates that plates coated with poly-L-lysine (PLL) exhibit an accelerated cell growth rate. When mesenchymal stem cells (MSCs) are cultured on PLL-coated substrates, it helps preserve their stemness properties and prevents senescence. PLL-coated plates promote enhanced MSC growth and up-regulation of genes associated with cell adhesion, differentiation, proliferation, and signaling. Furthermore, porous PLL-coated surfaces have been shown to enhance cell proliferation and support the osteogenic differentiation of MSCs.

Components

Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.

Caution

Sterile solutions are stable for up to 2 years when stored at 2-8°C. It should be stored desiccated at -20°C.

Analysis Note

Molecular weight based on viscosity. Also assayed by MALLS.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polylysine for skin regeneration: A review of recent advances and future perspectives
Payam Zarrintaj, et al.
Journal of Separation Science, 7(1) (2021)
Trial evaluation of bone marrow derived mesenchymal stem cells (MSCs) transplantation in revival of spermatogenesis in testicular torsion
Sabbaghi MA, et al.
Middle East Fertility Society journal, 17(4), 243-249 (2012)
Pre-B-cell colony-enhancing factor exerts a neuronal protection through its enzymatic activity and the reduction of mitochondrial dysfunction in in vitro ischemic models
Bi J, et al.
Journal of Neurochemistry, 120(2), 334-346 (2012)
Direct addition of poly-lysine or poly-ethylenimine to the medium: A simple alternative to plate pre-coating
Alexander Faussner, et al.
PLoS ONE, 17(7) (2022)
Aleksandra N Mladenovic Djordjevic et al.
Free radical biology & medicine, 162, 88-103 (2020-12-07)
Alzheimer's disease (AD) is the most common form of dementia worldwide, characterized by a progressive decline in a variety of cognitive and non-cognitive functions. The amyloid beta protein cascade hypothesis places the formation of amyloid beta protein aggregates on the

Articles

Poly-Lysine enhances cell binding with positively-charged surface ions, optimizing electrostatic interaction on culture surfaces for increased cell attachment.

Poly-Lysine enhances cell binding with positively-charged surface ions, optimizing electrostatic interaction on culture surfaces for increased cell attachment.

Poly-Lysine enhances cell binding with positively-charged surface ions, optimizing electrostatic interaction on culture surfaces for increased cell attachment.

Poly-Lysine enhances cell binding with positively-charged surface ions, optimizing electrostatic interaction on culture surfaces for increased cell attachment.

See All

Protocols

Adhere cells to solid substrates using poly-lysine, which enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service