Skip to Content
Merck

Skip To

P8908

Prostaglandin E1

synthetic, powder, BioReagent, suitable for cell culture

Synonym(s):

(11α,13E,15S)-11,15-Dihydroxy-9-oxoprost-13-enoic acid, Alprostadil, PGE1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48
UNSPSC Code:
12352209
NACRES:
NA.77
PubChem Substance ID:
EC Number:
212-017-2
Beilstein/REAXYS Number:
5294062
MDL number:
eCl@ss:
42020658
Biological source:
synthetic
Form:
powder
Assay:
≥98% (HPLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

product line

BioReagent

assay

≥98% (HPLC)

form

powder

potency

0.25-100 ng/mL

technique(s)

cell culture | mammalian: suitable

solubility

acetone: 10 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

O=C1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@H](CCCCC)O)[C@H](O)C1

InChI

1S/C20H34O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-17,19,21,23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+,19+/m0/s1

InChI key

GMVPRGQOIOIIMI-DWKJAMRDSA-N

General description

Prostaglandin E1 (PGE1) is a vasodilator.

Application

Prostaglandin E1 may be used in cell culture to protect human umbilical vein endothelial cells (HUVEC) from injury induced by hydrogen peroxide.

Biochem/physiol Actions

Prostaglandin E1 (PGE1) aids in the relaxation of corporal cavernosal tissue. It helps to preserve the patency of the ductus arteriosus. It is used to treat erectile dysfunction (ED).

Physical form

powder-0 °C; stock-frozen in working aliquots, avoid repeated freeze/thaw

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P7527P5515538903
biological source

synthetic

biological source

synthetic (organic)

biological source

synthetic

biological source

-

assay

≥98% (HPLC)

assay

≥60% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

product line

BioReagent

product line

BioXtra

product line

-

product line

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

form

powder

form

powder

form

(powder to crystals)

form

solid


Still not finding the right product?

Explore all of our products under Prostaglandin E1


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters