Skip to Content
Merck
All Photos(2)

Key Documents

P8791

Sigma-Aldrich

Polyglycine

mol wt 500-5,000

Synonym(s):

Glycine homopolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

form

lyophilized powder

Quality Level

mol wt

500-5,000

color

white to light yellow

mp

300 °C

storage temp.

−20°C

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Polyglycine has been used as an external intensity standard for measuring cross polarization (CP) nuclear magnetic resonance (NMR) spectrum measurements[1] and ion mobility mass spectrometry.[2] It is suitable for use as a model for UV resonance Raman spectroscopy studies.[3]

Biochem/physiol Actions

Polyglycine exists in two forms, namely the polyglycine I (PGI) with anti-parallel β-sheet structure and polyglycine II (PGII) with extended 31-helix. It is a most flexible polypeptide with minimal steric hindrance and its solubility increases in the presence of lithium ions.[3] It is present in mammals and plants. Polyglycine stretch associated with the chloroplast membrane protein, Toc5 is essential for envelop sorting.[4]

Preparation Note

Prepared by phosphorylation.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiran Kumar Morishetti et al.
Journal of the American Society for Mass Spectrometry, 21(4), 603-614 (2010-01-29)
The sequence and conformational effects on the gas-phase acidities of peptides have been studied by using two pairs of isomeric cysteine-polyglycine peptides, CysGly(3,4)NH(2) and Gly(3,4)CysNH(2). The extended Cooks kinetic method was employed to determine the gas-phase acidities using a triple
Solid-state 15 N NMR analysis of highly 15 N-enriched plant materials
Smernik RJ and Baldock JA
Plant and Soil, 275(1-2), 271-283 (2005)
Sergei Bykov et al.
The journal of physical chemistry. B, 114(19), 6636-6641 (2010-04-27)
Polyglycine (polygly) is an important model system for understanding the structural preferences of unfolded polypeptides in solution. We utilized UV resonance and visible Raman spectroscopy to investigate the conformational preferences of polygly peptides of different lengths in water containing LiCl
Koji Ikegami et al.
Cell structure and function, 35(1), 15-22 (2010-03-02)
Microtubules (MTs) play specialized roles in a wide variety of cellular events, e.g. molecular transport, cell motility, and cell division. Specialized MT architectures, such as bundles, axonemes, and centrioles, underlie the function. The specialized function and highly organized structure depend
Ogata, N., et al.
Polymer Journal, 8, 129-129 (1976)

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service