Skip to Content

P8787

o-Phenylenediamine dihydrochloride

chromogenic, tablet

Synonym(s):

1,2-Phenylenediamine tablet, OPD, OPD Tablet

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H4(NH2)2·2HCl
CAS Number:
Molecular Weight:
181.06
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Beilstein/REAXYS Number:
3912045
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

o-Phenylenediamine dihydrochloride, tablet, 4 mg substrate per tablet

Quality Segment

form

tablet

mp

258 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Nc1ccccc1N

InChI

1S/C6H8N2.2ClH/c7-5-3-1-2-4-6(5)8;;/h1-4H,7-8H2;2*1H

InChI key

RIIWUGSYXOBDMC-UHFFFAOYSA-N

General description

o-Phenylenediamine dihydrochloride (OPD) is a chromogenic substrate that can be used in enzyme-linked immunosorbent assay (ELISA) procedures as it utilizes horseradish peroxidase conjugates. The substrate produces a soluble end product that is orange-brown and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3N HCl or 3M H2SO4 and read at 492 nm.

Application

o-Phenylenediamine dihydrochloride has been used as a substrate for horseradish-peroxidase-conjugated secondary antibodies in enzyme-linked immunosorbent assay (ELISA).

Preparation Note

Dissolve one tablet in 0.05 M phosphate-citrate buffer, pH 5.0, to the desired concentration (typically an OPD concentration of 0.4 mg/ml is used). Add 40 μl of fresh 30% hydrogen peroxide per 100 ml of substrate buffer solution, immediately prior to use. Tablets are individually packaged in foil packets.


Still not finding the right product?

Explore all of our products under o-Phenylenediamine dihydrochloride


signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library