Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C11H6O3
CAS Number:
Molecular Weight:
186.16
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
200-639-7
MDL number:
Beilstein/REAXYS Number:
152784
Assay:
≥99% (HPLC)
Form:
powder
Product Name
Psoralen, ≥99% (HPLC)
Quality Segment
assay
≥99% (HPLC)
form
powder
fluorescence
λex 335 nm; λem 460 nm (pH 7.0)
storage temp.
2-8°C
SMILES string
O=C1Oc2cc3occc3cc2C=C1
InChI
1S/C11H6O3/c12-11-2-1-7-5-8-3-4-13-9(8)6-10(7)14-11/h1-6H
InChI key
ZCCUUQDIBDJBTK-UHFFFAOYSA-N
Application
Photochemical reagent for the investigation of nucleic acid structure and function.
Psoralen has been used:
- as a crosslinker for dsRNA for in- situ hybridization studies
- in testing melanin dispersal effects in fish scale melanophores
- to test its stimulatory effects in osteoblast a cell proliferation assay
- as a reference standard in high-performance liquid chromatography (HPLC)
Biochem/physiol Actions
Psoralen belongs to the linear type furanocoumarins. It intercalates and induces interstrand cross-links in DNA. On UV exposure psoralen is excited leading to irreversible intercalation with DNA by covalent bond formation. This property of psoralen ultraviolet A light (PUVA) is exploited in treating skin diseases and cutaneous T-cell lymphoma. However, usage psoralen also leads to hepatotoxicity and cytotoxicity by the generation of psoralen photoproducts (POPs). It may be useful in treating osteoporosis
Features and Benefits
This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
Still not finding the right product?
Explore all of our products under Psoralen
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Sens. 1
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
