Skip to Content
Merck

P8087

Monoclonal Anti-Plakoglobin (Catenin γ) antibody produced in mouse

clone 15F11, ascites fluid, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
15F11, monoclonal
Application:
ARR, ICC, IF, IHC (f), IP, WB
Citations:
16
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

15F11, monoclonal

form

buffered aqueous solution

mol wt

antigen 85 kDa

contains

15 mM sodium azide

species reactivity

bovine, canine, human

technique(s)

immunocytochemistry: suitable using cultured cells, immunohistochemistry (frozen sections): suitable, immunoprecipitation (IP): suitable, indirect immunofluorescence: 1:1,000 using cultured MDBK cells, microarray: suitable, western blot: 1:2,000 using cultured MDBK cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JUP(3728)

General description

Plakoglobin is a desmosomal protein that regulates developmental signalling in vertebrates. Tyrosine phosphorylation of plakoglobin has been linked to adenocarcinomas . Plakoglobin may function as a tumor suppressor in ovarian and breast cancers .

Immunogen

recombinant chicken plakoglobin.

Application

Monoclonal anti-plakoglobin (catenin γ) antibody is suitable for use in immunocytochemistry (using MDCK cells), immunoprecipitation, and in immunoblotting . The antibody may also be used for immunohistochemistry (frozen sections), microarray, western blot (1:500, using cultured MDBK cells) and indirect immunofluorescence (1:1000, using cultured MDBK cells).

Biochem/physiol Actions

The antibody recognizes the plakoglobin (catenin-γ) molecule (85 kDa and possibly a slightly lower band) by immunoblotting. It does not cross-react with β-catenin. The antibody reacts with plakoglobin in bovines, humans and dogs, but does not react with β catenin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library