Skip to Content
Merck
All Photos(1)

Key Documents

P7943

Sigma-Aldrich

L-α-Phosphatidylethanolamine from egg yolk

Type III, ≥97% (TLC), lyophilized powder

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphoethanolamine, L-α-Cephalin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

egg yolk

Quality Level

type

Type III

Assay

≥97% (TLC)

form

lyophilized powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[P](=O)(OC[C@H](OC(=O)CCCCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCC)(OCCN)O

InChI

1S/C40H80NO8P/c1-3-5-7-9-11-13-15-17-18-19-20-21-23-25-27-29-31-33-40(43)49-38(37-48-50(44,45)47-35-34-41)36-46-39(42)32-30-28-26-24-22-16-14-12-10-8-6-4-2/h38H,3-37,41H2,1-2H3,(H,44,45)/t38-/m1/s1

InChI key

NJGIRBISCGPRPF-KXQOOQHDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-α-Phosphatidylethanolamine from egg yolk has high unsaturated fatty acid content and elicits radical-scavenging activity.[1] L-α-Phosphatidylethanolamine (PE) is a membrane phospholipid and is interconverted to phosphotidylserine in the presence of phosphatidylserine synthase 2. PE is essential for maintaining the structural integrity of membranes.[2] It is the second most abundant phospholipid in animals, plants and yeast and the major lipid in bacteria. It is synthesized in mitochondria in mammals.[3]

Application

L-α-Phosphatidylethanolamine from egg yolk has been used:
  • as a component of lipid mixture for the synthesis of giant unilamellar vesicles (GUVs)[4]
  • in ceramide phosphoethanolamine (CPE) synthase assay using crude sphingomyelin synthase-related protein from Hela cells[5]
  • for the standard curve generation for the quantification of lipids in plantaris muscle homogenates from mice[6]

Packaging

Packaged under Argon.

Caution

Purity when prepared is 97-99%. Solutions exposed to room temperature can decompose 0.3-0.5% per day.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scolopendin 2, a cationic antimicrobial peptide from centipede, and its membrane-active mechanism
Lee H, et al.
Biochimica et Biophysica Acta - Biomembranes, 1848(2), 634-642 (2015)
Tingbo Ding et al.
Journal of lipid research, 56(3), 537-545 (2015-01-22)
Sphingomyelin synthase-related protein (SMSr) synthesizes the sphingomyelin analog ceramide phosphoethanolamine (CPE) in cells. Previous cell studies indicated that SMSr is involved in ceramide homeostasis and is crucial for cell function. To further examine SMSr function in vivo, we generated Smsr
Egg Yolk Phosphatidylethanolamine: Extraction Optimization, Antioxidative Activity, and Molecular Structure Profiling
Sun N, et al.
Journal of Food Science (2019)
Elevated whole muscle phosphatidylcholine: phosphatidylethanolamine ratio coincides with reduced SERCA activity in murine overloaded plantaris muscles
Fajardo VA, et al.
Lipids in Health and Disease, 17(1), 47-47 (2018)
All members in the sphingomyelin synthase gene family have ceramide phosphoethanolamine synthase activity
Ding T, et al.
Journal of Lipid Research, 56(3), 537-545 (2015)

Protocols

(soybean), ≥97%; L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder; L-α-Phosphatidylcholine, from egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder; Sphingomyelin, from chicken egg yolk, ≥95%

(soybean), ≥97%; L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder; L-α-Phosphatidylcholine, from egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder; Sphingomyelin, from chicken egg yolk, ≥95%

(soybean), ≥97%; L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder; L-α-Phosphatidylcholine, from egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder; Sphingomyelin, from chicken egg yolk, ≥95%

(soybean), ≥97%; L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder; L-α-Phosphatidylcholine, from egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder; Sphingomyelin, from chicken egg yolk, ≥95%

Questions

  1. What solvents are used to dissolve phosphatidylethanolamine?  What is the solubility?  What solvent is.least toxic to eucaryotic cells?

    1 answer
    1. This solubility is tested in CHCl3 at 25 mg/mL. Stock solutions in chloroform can be added to medium or buffer and allowed to stir prior to filtration. This allows the chloroform to evaporate off while keeping the Phosphatidylethanolamine in solution, thereby avoiding the risk of exposing cells to the solvent. An alternative solvent would be a mixture of chloroform and methanol, which would also be cytotoxic. Please see the sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/P7943-BULKSLCN2430.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service