Skip to Content
Merck
All Photos(1)

Key Documents

P7837

Sigma-Aldrich

Protein A from Staphylococcus aureus

Soluble, Cowan strain, recombinant, expressed in E. coli, aqueous solution, ≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

recombinant

expressed in E. coli

Quality Level

conjugate

unconjugated

Assay

≥95% (HPLC)

form

aqueous solution

mol wt

45,000

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

Staphylococcus aureus subsp. aureus NCTC 8325 ... SAOUHSC_00069(3919448)

Looking for similar products? Visit Product Comparison Guide

Application

Protein A may be conjugated with various reporter molecules including fluorescent dyes (FITC), enzyme markers (peroxidase, β-galactosidase, alkaline phosphatase), biotin, and colloidal gold without affecting the antibody binding site on the molecule. These conjugates are used to detect immunoglobulins in various immunochemical assays including western blotting, immunohistochemistry, and ELISA applications. In addition, protein A may be immobilized on a solid support such as agarose or acrylic beads for the purification of either polyclonal or monoclonal immunoglobulins. It is also routinely used for immunoprecipitation assays.

Biochem/physiol Actions

A highly stable cell surface receptor produced by several strains of Staphylococcus aureus.
Protein A is a highly stable cell surface receptor produced by several strains of Staphylococcus aureus. It consists of a single polypeptide chain of molecular weight 42 kDa, containing four repetitive domains rich in aspartic and glutamic acids but devoid of cysteine. It contains little or no carbohydrate and only 4 tyrosine residues and no tryptophans. Protein A is capable of binding to the Fc portion of immunoglobulins, especially IgGs, from a large number of species. One protein A molecule has been shown to bind at least 2 molecules of IgG simultaneously. The IgG binding domain of Protein A consists of three anti-parallel α-helicies, the third of which is disrupted when the protein is complexed with the Fc region of the immunoglobulins. Protein A will bind the Fc portion of human IgG subclasses, IgM, IgA and IgE; and mouse IgG1 (weakly), IgG2a, and IgG2b. Protein A also binds IgGs from other species, including monkey, rabbit, pig, guinea pig, dog, and cat.
Protein A also participates in a number of different protective biological functions including anti-tumor, toxic, and carcinogenic activities. In addition to acting as an immunomodulator, it also has antifungal and antiparasitic properties.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chaitali P Basole et al.
Cellular signalling, 40, 62-72 (2017-08-31)
The levels of cAMP are regulated by phosphodiesterase enzymes (PDEs), which are targets for the treatment of inflammatory disorders. We have previously shown that PDE8 regulates T cell motility. Here, for the first time, we report that PDE8A exerts part
Thierry Champion et al.
Methods in molecular biology (Clifton, N.J.), 988, 331-343 (2013-03-12)
Surface plasmon resonance (SPR) is a key technology to evaluate IgG effector functions in vitro involving binding to Fcgamma receptors (FcγR). We describe here the chemical coupling of protein A to a sensor chip. IgGs prepared either from purified antibodies
Irene Tom et al.
Current protocols in protein science, Chapter 27, Unit 27-Unit 27 (2013-04-03)
Functional protein microarrays offer the capability for high-throughput protein interaction analysis and have long promised to be a powerful tool for understanding protein interactions at the proteome scale. Although popular techniques for protein-protein interaction mapping like yeast-two-hybrid and affinity-purification mass
Microplate-based chromatin immunoprecipitation method, Matrix ChIP: a platform to study signaling of complex genomic events.
Flanagin S et al
Nucleic Acids Research, 36(3), e17-e17 (2008)
Sally L Sherwen et al.
Zoo biology, 34(4), 287-295 (2015-06-04)
Free range exhibits are becoming increasingly popular in zoos as a means to enhance interaction between visitors and animals. However very little research exists on the impacts of visitors on animal behaviour and stress in free range exhibits. We investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service