Skip to Content
Merck

P7174

Finnpipette® pipette tips

Finntip 1000, non-sterile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.01
UNSPSC Code:
41121600
Sterility:
non-sterile
Feature:
barrier: no
Packaging:
pack of 960 ea (Racks)
Manufacturer/tradename:
Finn 9401110
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

blue

sterility

non-sterile

feature

barrier: no

packaging

pack of 960 ea (Racks)

manufacturer/tradename

Finn 9401110

L

7.1 cm

maximum volume

1 mL

volume range

100-1000 μL

Features and Benefits

  • Enables accurate and precise pipetting
  • Designed for liquid handling with a uniform smooth surface
  • A hydrophobic surface prevents liquid retention
  • The rack packaging option has color-coding

Other Notes

Browse all pipette tips

Legal Information

Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Finnpipette® pipette tips




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library