Skip to Content
Merck

P6804

Microcapillary pipettes, positive displacement

volume range 1-10 μL

Synonym(s):

Positive Displacement Pipettes

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
Drummond Scientific 5-000-1001-X10
Material:
glass
Packaging:
pack of 100 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

glass

packaging

pack of 100 ea

manufacturer/tradename

Drummond Scientific 5-000-1001-X10

capacity

10 μL

volume range

1-10 μL

General description

Calibrated glass microcapillary pipettes with separate stainless steel plunger wires. Not damaged by dry sterilization above 400 °F (205 °C). Pipette 50% glycerol solutions accurately without aerosol formation. Single-use disposable pipette and plunger to avoid cross-contamination.

Application

Designed specifically for use in PCR work

Packaging

Package contains 100 pipettes, 100 plunger wires, and one reusable ring-style handle.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library