Skip to Content
Merck
All Photos(1)

Key Documents

P6649

Millipore

Protein A–Sepharose 6MB

aqueous ethanol suspension

Synonym(s):

Protein A–Agarose macrobeads

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56
Pricing and availability is not currently available.

form

aqueous ethanol suspension

Quality Level

extent of labeling

~1 mg per mL

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

capacity

~6 mg/mL binding capacity (human IgG)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

P6649-10ml′s updated product number is GE17-0469-01

Application

Protein A-Sepharose is used for affinity chromatography, antibody purification and characterization, immunoaffinity matrices, protein chromatography, protein A, G and L resins, and recombinant protein expression and analysis. Protein A-Sepharose has been used to develop strategies for investigating protein interactions, to improve the detection of celiac disease, and to study diabetes in children.
Protein A–Sepharose 6MB has been used in immunoprecipitation.[1][2][3]

Physical form

Suspension in 20% ethanol

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F

Flash Point(C)

38 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L F Wang et al.
Gene, 169(1), 53-58 (1996-02-22)
Epitope tagging (Eta) is becoming an increasingly useful technique in molecular biology and biotechnology for the detection, characterisation and purification of recombinant proteins (re-proteins). Here we describe a novel Eta system composed of two different monoclonal antibodies (mAb; D11 and
Sundar Rajan Selvaraj et al.
Molecular biology of the cell, 19(12), 5579-5592 (2008-09-26)
Retinol-binding protein (RBP) is secreted out of the cell in its ligand-bound holo-form. The apo-form of RBP is selectively retained within the endoplasmic reticulum (ER) by a mechanism that remains unknown. Using isolated microsomal system, we have recapitulated the biogenesis
G V Hillyer et al.
The American journal of tropical medicine and hygiene, 38(3), 547-552 (1988-05-01)
Total RNA containing messenger RNA has been isolated from adult Fasciola hepatica and translated in vitro using the rabbit reticulocyte lysate system. L-[35S]-methionine labeled translation products have been immunoprecipitated with sera collected from rabbits infected with F. hepatica, rabbits immunized
Gaurav Dugar et al.
Molecular cell, 69(5), 893-905 (2018-03-03)
Cas9 nucleases naturally utilize CRISPR RNAs (crRNAs) to silence foreign double-stranded DNA. While recent work has shown that some Cas9 nucleases can also target RNA, RNA recognition has required nuclease modifications or accessory factors. Here, we show that the Campylobacter
Nikolai Peschek et al.
The EMBO journal, 38(16), e101650-e101650 (2019-07-18)
Small regulatory RNAs (sRNAs) are crucial components of many stress response systems. The envelope stress response (ESR) of Gram-negative bacteria is a paradigm for sRNA-mediated stress management and involves, among other factors, the alternative sigma factor E (σE ) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service