Skip to Content
Merck

P6499

PNU-282987 hydrate

solid, ≥98% (HPLC)

Synonym(s):

N-(3R)-1-Azabicyclo[2.2.2]oct-3-yl-4-chloro-benzamide monohydrochloride hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H17ClN2O · HCl · xH2O
Molecular Weight:
301.21 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Cl[H].[H]O[H].Clc1ccc(cc1)C(=O)N[C@H]2CN3CCC2CC3

InChI

1S/C14H17ClN2O.ClH.H2O/c15-12-3-1-11(2-4-12)14(18)16-13-9-17-7-5-10(13)6-8-17;;/h1-4,10,13H,5-9H2,(H,16,18);1H;1H2/t13-;;/m0../s1

InChI key

OCWLMMBVXIESNP-GXKRWWSZSA-N

Application

PNU-282987 hydrate has been used to check its activity in wound repair by inhibiting AGE (advanced glycation end products)-mediated tumor necrosis factor-α (TNF-α) production in a streptozotocin (STZ)-induced diabetic mouse model. It has also been used to evaluate the effects of nicotinic α-7 acetylcholine receptor (nAChRα7) activation on non-diabetic wound healing.

Biochem/physiol Actions

PNU-282987 is an agonist of nicotinic α-7 acetylcholine receptor (nAChRα7). It helps to decrease acute lung injury (ALI), stimulated by lipopolysaccharide (LPS). PNU-282987 can increase GABAergic synaptic activity in brain slices and helps to bring back auditory gating deficits in anesthetized rats.
Decreased expression of a homomeric alpha7 nicotinic acetylcholine receptor (nAChR) is connected with inability to process sensory information in schizophrenia. PNU-282987 is a novel selective agonist of the alpha7 nAChR that evoked whole-cell currents from cultured rat hippocampal neurons that were sensitive to the selective alpha7 nAChR antagonist methyllycaconitine (MLA) and enhanced GABAergic synaptic activity. The alpha7 nAChR agonist PNU-282987 improves auditory gating and enhances hippocampal oscillatory activity. These results provide further support for the concept that drugs that selectively activate alpha7 nAChRs may offer a novel, potential pharmacotherapy in treatment of schizophrenia.
Selective α7 nicotinic receptor agonist. K= 27 nM; displays negligible blockade of α1β1γδ and α3β4 nAChRs (IC50 = 60 mM); enhances GABAergic synaptic activity in the hippocampus in vitro and enhances amphetamine-induced hippocampal theta wave activity.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Disclaimer

Air sensitive


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library