Skip to Content
Merck
All Photos(1)

Key Documents

P6181

Sigma-Aldrich

Endoproteinase Glu-C from Staphylococcus aureus V8

suitable for protein sequencing, lyophilized powder

Synonym(s):

V8 Protease

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

grade

Proteomics Grade

Quality Level

form

lyophilized powder

analyte chemical class(es)

amino acids

suitability

suitable for protein sequencing

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Endoproteinase Glu-C from Staphylococcus aureus V8 has been used for:
  • obtaining proteolytic cleavage fragments of the S-layer protein (from Bacillus stearothermophilus ATCC 12980) to perform affinity studies.
  • the enzymatic cleavage of native VSTx-3 (voltage sensor toxin 3) peptide for its sequence determination.
  • limited proteolysis of recombinant purified PimA (phosphatidylinositol mannosyltransferase).
  • the digestion of glycosylated hemoglobin for isotope dilution liquid chromatography-tandem mass spectrometry analysis.

Biochem/physiol Actions

Endoproteinase Glu-C from Staphylococcus aureus strain V8 is a serine endoprotease, which hydrolyzes peptide bonds at the carboxyl side of glutamyl and aspartyl residues. The specificity of Glu-C is dependent upon the buffer and pH employed as well as the structure around the potential cleavage site. In ammonium acetate (pH 4.0) or ammonium bicarbonate (pH 7.8), the enzyme preferentially cleaves glutamyl bonds; whereas, in phosphate buffer (pH 7.8) Glu-C will cleave at either site. Glu-C is reported to be active in the presence of 0.2% SDS (sodium dodecyl sulfate) and in 4.0M urea.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Pietropaolo et al.
The Journal of general virology, 80 ( Pt 7), 1807-1816 (1999-07-28)
Annexin II has been identified as a human cytomegalovirus (HCMV)-binding protein, shown to be a component of purified virions and proposed as a cellular receptor for the virus. In addition, annexin II is capable of associating with the major HCMV
Q Xu et al.
The Biochemical journal, 341 ( Pt 3), 733-737 (1999-07-27)
A galactose-binding lectin isolated from the venom of Trimeresurus stejnegeri is a homodimer C-type lectin. The cloned cDNA encoding the monomer of Trimeresurus stejnegeri lectin (TSL) was sequenced and found to contain a 5'-end non-coding region, a sequence which encodes
Daniel B Williamson et al.
The Journal of biological chemistry, 297(3), 101055-101055 (2021-08-20)
Fibrillin-1 (FBN1) is the major component of extracellular matrix microfibrils, which are required for proper development of elastic tissues, including the heart and lungs. Through protein-protein interactions with latent transforming growth factor (TGF) β-binding protein 1 (LTBP1), microfibrils regulate TGF-β
Secondary structure reshuffling modulates glycosyltransferase function at the membrane.
Giganti D et al.
Nature Chemical Biology, 11, 16-16 (2015)
M Malkoski et al.
Antimicrobial agents and chemotherapy, 45(8), 2309-2315 (2001-07-14)
Caseinomacropeptide (CMP) is a heterogeneous C-terminal fragment (residues 106 to 169) of bovine milk kappa-casein composed of glycosylated and phosphorylated forms of different genetic variants. We have demonstrated that CMP has growth-inhibitory activity against the oral opportunistic pathogens Streptococcus mutans

Protocols

An optimized LC-MS/MS based workflow for low artifact tryptic digestion and peptide mapping of monoclonal antibody, adalimumab (Humira) using filter assisted sample preparation (FASP).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service