Skip to Content
Merck

P4634

TWEEN® 85

Synonym(s):

Polyoxyethylenesorbitan Trioleate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12161900
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Segment

assay

≥50%

form

viscous liquid

composition

Oleic acid, ≥50% (balance primarily C10-C18 fatty acids)

concentration

≥50% (GC)

density

1,03 g/cm3 at 20 °C

General description

Tween 85 is a polyoxyethylene sorbitan trioleate non-ionic surfactant.

Application

Tween 85 has been used in a study to assess the potential application of niosomes for the delivery of ammonium glycyrrhizinate for the treatment of inflammatory diseases. It has also been used in a study to investigate the joint-action of antifouling substances in copper-free paints.

Legal Information

TWEEN is a registered trademark of Croda International PLC


Still not finding the right product?

Explore all of our products under TWEEN® 85


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

>300.2 °F - DIN 51758

flash_point_c

> 149 °C - DIN 51758

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library