Skip to Content
Merck

P4380

Protamine sulfate

powder

Synonym(s):

Salmine, Salmine from salmon

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Protamine sulfate salt from salmon, Grade II

biological source

fish (salmon)

Quality Segment

type

Grade II

form

powder

color

white to off-white

solubility

6 M HCl: 25 mg/mL, clear, colorless to light yellow

General description

Protamine sulfate (PS), a polycationic, positively charged protein is mostly made up of poly(l-arginine). It is obtained from salmon sperm protein.

Application

Protamine sulfate salt from salmon has been used in the generation of protamine silica nanocomposites.
May be used for the removal of DNA from protein samples or for the purification of DNA binding proteins. It may be used as an alternative to polybrene in retroviral-mediated gene transfer.

Protamine in the form of solid lipid nanoparticles (SLN) promoted transfection with plasmid DNA more efficiently and with less cytotoxicity than comparable SLNs composed of Esterquat-1.
Protamine in the form of solid lipid nanoparticles (SLN) promoted transfection with plasmid DNA more efficiently and with less cytotoxicity than comparable SLNs composed of Esterquat-1.

Biochem/physiol Actions

Protamine sulfate is highly preferred in cardiovascular surgical procedures to reverse the anticoagulant properties of heparin. It is used for biomedical and biotechnological applications due to its biodegradablity.
Protamine sulfate is a small cationic protein that binds and precipitates DNA. Neutralizes the anticoagulant activity of heparin. Inhibits lipoprotein lipase.

Analysis Note

May contain insolubles as well as histones.


Still not finding the right product?

Explore all of our products under Protamine sulfate salt from salmon


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
P4380-100G04061832948430
P4380-25G04061834374435