Skip to Content
Merck
All Photos(3)

Key Documents

P4334

Sigma-Aldrich

Anti-Protein Kinase Cα antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-PKCα

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen 80 kDa

contains

15 mM sodium azide

species reactivity

mouse, rat

technique(s)

microarray: suitable
western blot: 1:20,000 using mouse NIH 3T3 fibroblast lysate
western blot: 1:50,000 using rat brain extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Prkca(18750)
rat ... Prkca(24680)

General description

Anti-Protein Kinase Cα is produced in rabbit using a synthetic peptide conjugated to keyhole limpet hemocyanin (KLH) as the immunogen. PKC is a phospholipid dependent enzyme, activated by the lipid 1,2-diacylglycerol (DAG), an intracellular second messenger produced from hydrolysis of inositol phospholipids, in response to a variety of hormones, growth factors, and neurotransmitters.
PKC α is a conventional isoform of Protein Kinase C (76-93 kD) and a member of serine/threonine (Ser/Thr) protein kinases. It is expressed ubiquitously in tissues and is the major isoform of PKC in fibroblasts. Stimulation and overexpression of PKC α increases the growth rate of cells in culture. Anti-Protein Kinase C α antibody is useful for studying expression of PKC α in normal and neoplastic tissues. It is also useful for detecting PKC α isoenzyme in brain tissue and cell extracts. Furthermore, it can be used in chemiluminescence detection systems to detect PKC α. Rabbit anti-protein kinase c α antibody reacts specifically with PKC α (80 kD) of rat brain extract or NIH3T3 mouse fibroblast lysate.

Specificity

The antibody reacts in immunoblotting with PKCα in rat brain extract. Staining is specifically inhibited with PKCα immunizing peptide (659-672).

Immunogen

Synthetic peptide corresponding to amino acids from the C-terminal variable (V5) region of rat PKC α conjugated to KLH.

Application

Anti-Protein Kinase C α has been used in immunoblotting and immunohistochemistry.
Anti-Protein Kinase Cα antibody produced in rabbit has been used in light microscopic—immunocytochemical analysis and immunofluorescent labelling.

Biochem/physiol Actions

Protein Kinase C (PKC) plays a crucial role in signal transduction leading to cellular regulation, cell growth and differentiation, oncogenesis and modulation of neurotransmission. PKC is a phospholipid dependent enzyme, activated by the lipid 1,2-diacylglycerol (DAG), an intracellular second messenger produced as a result from hydrolysis of inositol phospholipids, in response to a variety of hormones, growth factors and neurotransmitters. PKC is also the major cellular receptor for the tumor promoting phorbol esters. PKC action is mediated by binding to specific receptors for activated C-kinase (RACKs) and through the phosphorylation of several cellular substrates. Overexpression and stimulation of PKC α leads to enhanced growth rate of cells in culture. Anti-Protein Kinase C α antibody is useful for studying expression of PKC α in normal and neoplastic tissues.

Physical form

Supplied as a liquid containing 15 mM sodium azide as preservative

Storage and Stability

For continuous use, store at 2-8 °C. For extended storage freeze in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mu Qiao et al.
Frontiers in molecular neuroscience, 8, 81-81 (2016-01-19)
Understanding how the nervous system functions requires mapping synaptic connections between neurons. Several methods are available for imaging neurons connected by chemical synapses, but few enable marking neurons connected by electrical synapses. Here, we demonstrate that a peptide transporter, Pept2
Koen Jansen et al.
Neuropeptides, 37(1), 57-65 (2003-03-15)
The suprachiasmatic nucleus (SCN) is the locus of the main pacemaker for circadian behavioral rhythms. In common voles, variation in circadian behavioral rhythmicity correlates with vasopressin (AVP) immunoreactive cells in the SCN. Here we studied the immunostaining of four AVP
Cone photoreceptors and potential UV vision in a subterranean insectivore, the European mole
Glosmann M et al
Journal of Vision, 8, 1-12 (2008)
Protein kinase C: structural and spatial regulation by phosphorylation, cofactors, and macromolecular interactions
Newton AC
Chemical Reviews, 101(8), 2353-2364 (2001)
K Almholt et al.
The Biochemical journal, 337 ( Pt 2), 211-218 (1999-01-12)
The alpha isoform of protein kinase C (PKCalpha) is a ubiquitous protein kinase, which, upon activation, translocates rapidly from the cytoplasm to the plasma membrane. To follow this translocation, PKCalpha was tagged with a highly fluorescent derivative of green fluorescent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service