Skip to Content
Merck

P4240

1,2-Dipalmitoylphosphatidylinositol 3,4,5-trisphosphate tetrasodium salt

≥98%

Synonym(s):

1-(1,2-Dihexadecanoylphosphatidyl)inositol-3,4,5-trisphosphate tetrasodium salt, PtdIns-(3,4,5)-P3 (1,2-Dipalmitoyl) tetrasodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C41H78Na4P4O22
Molecular Weight:
1138.90
NACRES:
NA.77
UNSPSC Code:
12352207
Assay:
≥98%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

powder

storage temp.

−20°C

Biochem/physiol Actions

Proposed as signal transduction modulator.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library