Skip to Content
Merck
All Photos(1)

Documents

P4236

Sigma-Aldrich

Cyclic Pifithrin-α hydrobromide

≥98% (HPLC)

Synonym(s):

2-(4-Methylphenyl)imidazo[2,1-b]-5,6,7,8-tetrahydrobenzothiazole hydrobromide, Cyclic PFT-α hydrobromide, Pifithrin-α, cyclic, QB102

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2S · HBr
CAS Number:
Molecular Weight:
349.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: 20 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

Br[H].Cc1ccc(cc1)-c2cn3c4CCCCc4sc3n2

InChI

1S/C16H16N2S.BrH/c1-11-6-8-12(9-7-11)13-10-18-14-4-2-3-5-15(14)19-16(18)17-13;/h6-10H,2-5H2,1H3;1H

InChI key

SGNCOAOESGSEOP-UHFFFAOYSA-N

Application

Cyclic Pifithrin-α hydrobromide has been used as p53 inhibitor to study its role in cigarette smoke−induced apoptosis of pulmonary endothelial cells.

Biochem/physiol Actions

A stable analog of Pifithrin-α (Product Code P4359) with similar biological activities and lower cellular toxicity.

Packaging

Protect from light.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

p53 mediates cigarette smoke-induced apoptosis of pulmonary endothelial cells: inhibitory effects of macrophage migration inhibitor factor
Damico R, et al.
American Journal of Respiratory Cell and Molecular Biology, 44(3), 323-332 (2011)
Pifithrin-alpha inhibits p53 signaling after interaction of the tumor suppressor protein with hsp90 and its nuclear translocation
Murphy P J M, et al.
The Journal of Biological Chemistry, 279(29), 30195-30201 (2004)
Rachel Damico et al.
American journal of respiratory cell and molecular biology, 44(3), 323-332 (2010-05-08)
Exposure to cigarette smoke (CS) is the most common cause of emphysema, a debilitating pulmonary disease histopathologically characterized by the irreversible destruction of lung architecture. Mounting evidence links enhanced endothelial apoptosis causally to the development of emphysema. However, the molecular
Tatiana Cardinal et al.
PLoS genetics, 16(9), e1009008-e1009008 (2020-09-09)
Hirschsprung disease (HSCR) is a complex genetic disorder of neural crest development resulting in incomplete formation of the enteric nervous system (ENS). This life-threatening neurocristopathy affects 1/5000 live births, with a currently unexplained male-biased ratio. To address this lack of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service