Skip to Content
Merck

P4020

Protamine sulfate

amorphous powder

Synonym(s):

Salmine, Salmine from salmon

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Protamine sulfate salt from salmon, Grade X, amorphous powder

type

Grade X

Quality Segment

form

amorphous powder

impurities

histone, free (Millon test)

color

white to off-white

solubility

6 M HCl: 25 mg/mL

General description

Protamine are proteins rich in cysteine and arginine. It possesses many phosphorylation sites. It is found to be localized to the sperm head.

Application

Protamine sulfate salt from salmon has been used:
  • in the pretreament of cells to study its effect on respiratory syncytial virus infection
  • in lentivirus production
  • to derivatize it into a diazeniumdiolate NO donor

May be used for the removal of DNA from protein samples or for the purification of DNA binding proteins. It may be used as an alternative to polybrene in retroviral-mediated gene transfer.

Protamine in the form of solid lipid nanoparticles (SLN) promoted transfection with plasmid DNA more efficiently and with less cytotoxicity than comparable SLNs composed of Esterquat-1.
Protamine in the form of solid lipid nanoparticles (SLN) promoted transfection with plasmid DNA more efficiently and with less cytotoxicity than comparable SLNs composed of Esterquat-1.

Biochem/physiol Actions

Protamine sulfate is a small cationic protein that binds and precipitates DNA. Neutralizes the anticoagulant activity of heparin. Inhibits lipoprotein lipase.


Still not finding the right product?

Explore all of our products under Protamine sulfate salt from salmon


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
P4020-10G04061832941301
P4020-25G04061834373445
P4020-5G04061834373452
P4020-100G04061834373421
P4020-1G04061834373438