Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C24H16N2O2
CAS Number:
Molecular Weight:
364.40
UNSPSC Code:
41116105
NACRES:
NA.32
PubChem Substance ID:
EC Number:
217-304-6
Beilstein/REAXYS Number:
325741
MDL number:
product line
BioReagent
Quality Segment
form
powder
mp
242-246 °C (lit.)
suitability
suitable for scintillation
SMILES string
c1ccc(cc1)-c2cnc(o2)-c3ccc(cc3)-c4ncc(o4)-c5ccccc5
InChI
1S/C24H16N2O2/c1-3-7-17(8-4-1)21-15-25-23(27-21)19-11-13-20(14-12-19)24-26-16-22(28-24)18-9-5-2-6-10-18/h1-16H
InChI key
MASVCBBIUQRUKL-UHFFFAOYSA-N
General description
1,4-Bis(5-phenyl-2-oxazolyl) benzene, also known as POPOP, is a well-known scintillation luminophore. It is an efficient fluorescent organic compound with high stokes shift. The POPOP dye is photostable in polar protic and polar aprotic solvents. The resulting photochemical quantum yield and the photodecomposition rate depend on the solvent′s electron affinity.
Application
1,4-Bis(5-phenyl-2-oxazolyl) benzene or POPOP is widely used to detect radioactivity by scintillation counting.
Features and Benefits
1,4-Bis(5-phenyl-2-oxazolyl) benzene or POPOP has the following advantages:
- High fluorescence quantum yield.
- High molar absorptivity.
- Large Stokes Shift.
- High Photostability in most organic solvents.
Still not finding the right product?
Explore all of our products under 1,4-Bis(5-phenyl-2-oxazolyl)benzene
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
