Skip to Content

P3512

Stearyl palmitate

≥99%

Synonym(s):

Palmitic acid stearyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H68O2
CAS Number:
Molecular Weight:
508.90
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
EC Number:
220-000-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant (vegetable)

Quality Segment

assay

≥99%

form

powder

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

InChI

1S/C34H68O2/c1-3-5-7-9-11-13-15-17-18-19-21-23-25-27-29-31-33-36-34(35)32-30-28-26-24-22-20-16-14-12-10-8-6-4-2/h3-33H2,1-2H3

InChI key

BILPUZXRUDPOOF-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Stearyl palmitate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library