Skip to Content
Merck
All Photos(1)

Key Documents

P3164

Sigma-Aldrich

Anti-Pig Serum antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Serum, pig

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

pig

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Guinea pig serum is highly enriched with L-asparaginase activity. This L-asparaginase activity imparts inhibitory or anti-lymphoma effects to pig serum.

Specificity

Strong reactivity with normal pig serum has been determined.

Application

Anti-Pig Serum antibody produced in rabbit has been used in differential staining.
Anti-Pig Serum antibody produced in rabbit has been used in the dual differential staining of blastocysts to assess the number of inner cell mass (ICM) cells and trophectoderm (TE) cells.

Quality

The antiserum has been treated to remove lipoproteins.

Physical form

Rabbit Anti-Pig Serum is provided as a liquid containing 0.1% sodium azide as preservative.

Storage and Stability

For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Lin et al.
Reproductive toxicology (Elmsford, N.Y.), 75, 96-109 (2017-12-17)
In this study, we used a pig model to investigate the effects of α-solanine (a natural toxin found mainly in potato sprouts) on oocyte maturation, quality and subsequent embryonic development. We found that α-solanine (10 μM) disturbed meiotic resumption and increased
Evidence that the L-Asparaginase Activity of Guinea Pig Serum is responsible for its Antilymphoma Effects
Broome JD
Nature, 191, 1114-1115 (1961)
Potential functional roles of follistatin on bovine somatic cell nuclear transfer embryos
Lee KB, et al.
Korean Journal of Agricultural Science, 40(4), 353-358 (2013)
Ling Sun et al.
Journal of animal science and technology, 66(4), 726-739 (2024-08-21)
This study was conducted to investigate whether lysophosphatidic acid (LPA) could improve the development of porcine somatic cell nuclear transfer (SCNT) embryos. Porcine SCNT-derived embryos were cultured in chemically defined polyvinyl alcohol (PVA)-based porcine zygote medium (PZM)-4 without or with
alpha-Solanine impairs oocyte maturation and quality by inducing autophagy and apoptosis and changing histone modifications in a pig model.
Lin T, et al.
Reproductive Toxicology, 75, 96-109 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service