Skip to Content
Merck

P3069

Poly-L-lysine–FITC Labeled

mol wt 30,000-70,000

Synonym(s):

FITC-Labeled Poly-ʟ-lysine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352209
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

mol wt

precursor poly-L-lysine • HBr 30,000-70,000, 30,000-70,000

extent of labeling

0.003-0.01 mol FITC per mol lysine monomer

color

yellow to orange

application(s)

cell analysis

storage temp.

−20°C

Application


  • Application of fluorescence recovery after photobleaching to diffusion of a polyelectrolyte in a multilayer film.: This study utilizes FITC-labeled Poly-L-lysine to investigate the diffusion dynamics of polyelectrolytes in multilayer films via fluorescence recovery after photobleaching, providing insights into molecular interactions and film assembly processes (Picart C et al., 2005).

  • Detection of charges and their distribution on dialysis membranes with cationic/anionic dyes using confocal laser scanning microscopy.: This research demonstrates the use of FITC-labeled Poly-L-lysine among other dyes to detect and analyze the distribution of charges on dialysis membranes, which is crucial for improving the performance and safety of dialysis treatments (Thomaneck U et al., 1991).

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.


Still not finding the right product?

Explore all of our products under Poly-L-lysine–FITC Labeled


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library