Skip to Content

P2499

PD 173074

≥96% (HPLC), FGFR1 and receptor-3 inhibitor, powder

Synonym(s):

N-[2-[[4-(Diethylamino)butyl]amino-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl]-N′-(1,1-dimethylethyl)urea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H41N7O3
CAS Number:
Molecular Weight:
523.67
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥96% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

PD 173074, ≥96% (HPLC), powder

Quality Segment

assay

≥96% (HPLC)

form

powder

color

faintly yellow to dark yellow

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCN(CC)CCCCNc1ncc2cc(c(NC(=O)NC(C)(C)C)nc2n1)-c3cc(OC)cc(OC)c3

InChI

1S/C28H41N7O3/c1-8-35(9-2)13-11-10-12-29-26-30-18-20-16-23(19-14-21(37-6)17-22(15-19)38-7)25(31-24(20)32-26)33-27(36)34-28(3,4)5/h14-18H,8-13H2,1-7H3,(H3,29,30,31,32,33,34,36)

InChI key

DXCUKNQANPLTEJ-UHFFFAOYSA-N

Gene Information

human ... FGFR1(2260), KDR(3791)
mouse ... Pdgfrb(18596)

Application

PD 173074 has been used:
  • to treat epithelial-mesenchymal transition (EMT)-induced cell lines in order to study its therapeutic use for head and neck squamous cell carcinoma (HNSCC)
  • in combination with PD0325901 to test whether an alternative downstream pathway for (fibroblast growth factor) FGF signalling is adopted during human hypoblast induction
  • to study its potential role in acute lymphoblastic leukaemia (ALL)-derived cell lines (TOM-1 and NALM-20)

Biochem/physiol Actions

PD 173074 inhibitor is specific for receptor tyrosine kinases. It competes with adenosine triphosphate (ATP) for its inhibition. PD 173074 prevents the FGF/ vascular endothelial growth factor (VEGF) induced angiogenesis.
PD 173074 is a fibroblast growth factor receptor 3 (FGFR3) inhibitor: IC50 = 5 nM inhibition of FGFR3 autophosphorylation. PD 173074 arrests the G0/G1 phase of FGFR3-expressing cells. It is 100-fold more selective for FGFR3 than for VEGF receptors, IGF-1 receptors, and MAPKs.
PD 173074 is a fibroblast growth factor receptor 3 inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Still not finding the right product?

Explore all of our products under PD 173074


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library