Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C29H42N7O17P3S
CAS Number:
Molecular Weight:
885.67
NACRES:
NA.51
PubChem Substance ID:
UNSPSC Code:
41106305
MDL number:
Assay:
~95%
Form:
powder
assay
~95%
Quality Segment
form
powder
storage temp.
−20°C
SMILES string
[Li].CC(C)(COP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1OP(O)(O)=O)n2cnc3c(N)ncnc23)C(O)C(=O)NCCC(=O)NCCSC(=O)Cc4ccccc4
InChI
1S/C29H42N7O17P3S.Li.H/c1-29(2,24(40)27(41)32-9-8-19(37)31-10-11-57-20(38)12-17-6-4-3-5-7-17)14-50-56(47,48)53-55(45,46)49-13-18-23(52-54(42,43)44)22(39)28(51-18)36-16-35-21-25(30)33-15-34-26(21)36;;/h3-7,15-16,18,22-24,28,39-40H,8-14H2,1-2H3,(H,31,37)(H,32,41)(H,45,46)(H,47,48)(H2,30,33,34)(H2,42,43,44);;
InChI key
BYSZOQQABOXIQX-UHFFFAOYSA-N
Application
Coenzyme A functions as an acyl group carrier, acetyl-CoA. Phenylacetyl coenzyme A (Phenylacetyl-CoA) is used to study the mechanism of action of the TetR family transcriptional repressor PaaR. Phenylacetyl-CoA is used to study the phenylacetyl-CoA monooxygenase complex found in bacteria such as E. coli.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P2153-5MG | 04061833264621 |