Skip to Content
Merck
All Photos(1)

Key Documents

P2144

Sigma-Aldrich

Peanut oil

delivery vehicle for lipophilic compounds

Synonym(s):

Arachis Oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

peanut oil

form

liquid

composition

Unsaturated fatty acids, ~80%

density

0.91 g/mL at 25 °C (lit.)

functional group

oleic acid

lipid type

oils

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Peanut oil is extracted from Arachis hypogaea L, is a source of edible oil in the tropical region. Peanut oil has high concentration of oleic and linoleic acid. It has many industrial applications like soap, cosmetics etc.

Application

Non-polar solvent, largely consisiting of long-chain glycerides of fatty acids, used for the delivery of lipophilic compounds.
Peanut oil has been used in the preparation of tamoxifen stock solution. It has also been used for dissolving the pesticide and fungicide compounds for in vivo study.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The combined antiandrogenic effects of five commonly used pesticides
Birkhoj M, et al.
Toxicology and Applied Pharmacology, 201(1), 10-20 (2004)
Inducible gene targeting in the neonatal vasculature and analysis of retinal angiogenesis in mice
Pitulescu ME, et al.
Nature Protocols, 5(9), 1518-1518 (2010)
Chemical analyses of groundnut (Arachis hypogaea) oil
Anyasor GN, et al.
Pakistan journal of nutrition, 8(3), 269-272 (2009)
Mechanisms of action underlying the antiandrogenic effects of the fungicide prochloraz
Laier P, et al.
Toxicology and Applied Pharmacology, 213(2), 160-171 (2006)
Sergio Parra et al.
BMC pharmacology & toxicology, 15, 29-29 (2014-06-06)
Regulator of G protein signaling (RGS) proteins suppress G protein coupled receptor signaling by catalyzing the hydrolysis of Gα-bound guanine nucleotide triphosphate. Transgenic mice in which RGS-mediated regulation of Gαi2 is lost (RGS insensitive Gαi2G184S) exhibit beneficial (protection against ischemic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service