Skip to Content

P1874

Anti-Huntingtin Disease (HD/HTT) Antibody

mouse monoclonal, 3B5H10

Synonym(s):

Anti-Huntingtin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
3B5H10, monoclonal
Application:
ICC, IP, WB
Citations:
45
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-Polyglutamines antibody, Mouse monoclonal, ~2 mg/mL, clone 3B5H10, purified from hybridoma cell culture

biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

3B5H10, monoclonal

form

buffered aqueous solution

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: 1-2 μg/mL using extract of HEK-293T cells transfected with an N-terminal 171 amino acid fragment of human Huntingtin with a 68 glutamine stretch

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTT(3064)

General description

Abnormal stretch of triplet CAG (or CAA) codon produces a homomeric repeat of glutamine residues beyond the critical threshold and will lead to neurodegenerative disease. Monoclonal mouse anti-polyglutamine antibody can be used to detect poly Q protein expression and in western blot. Monoclonal anti-polyglutamines antibody reacts specifically to homomeric polyglutamines independent of the species.

Immunogen

GST-human Huntingtin (N-terminal fragment of 171 amino acids containing 65Q).

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Monoclonal anti-polyglutamines antibody may be used for ELISA, immunoblot and slot blot. It can also be used in immunoprecipitation and immunocytochemistry.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Legal Information

This product is for in vitro use only. It is not to be used for commercial purposes. Use of this product to produce products for sale or for diagnostic, therapeutic or drug discovery purposes is prohibited. In order to obtain a license to use this product for commercial purposes, contact the Regents of the University of California. This product is sold under license to U.S. Patent No. 6,291,652, owned by the Regents of the University of California.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library