Skip to Content
Merck
All Photos(2)

Documents

P1867

Sigma-Aldrich

Plasmin from human plasma

lyophilized powder, ≥2.0 units/mg protein

Synonym(s):

Fibrinolysin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42030239
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥2.0 units/mg protein

impurities

HIV, Hepatitis B and Hepatitis C, tested negative

UniProt accession no.

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Gene Information

human ... PLG(5340)

Looking for similar products? Visit Product Comparison Guide

General description

Plasmin functions as a key enzyme of the fibrinolytic cascade, and is also important in inflammation processes.

Application

A complex between plasmin and an inhibitor has been isolated in a study via affinity chromatography from urokinase-activated human plasma. It has also been used in a study to investigate activation of human epithelial sodium channel (ENaC) by plasmin and chymotrypsin.

Biochem/physiol Actions

It is inhibited by α 2-antiplasmin and its interaction with fibrin is blocked. It digests the N-terminal region of the cytoplasmic protein αsynuclein preventing its uptake by surrounding cells.
Plasmin exhibits preferential cleavage at the carboxyl side of lysine and arginine residues with higher selectivity than trypsin. Converts polymerized fibrin into soluble products.
pH Optimum: 8.5
pH 7.5: ~40% of maximal activity, pH 9.5: ~50% of maximal activity
Temperature Optimum: 37 °C with rapid inactivation at 56 °C

Packaging

Package size based on protein content

Unit Definition

One unit will produce one μmole of p-Nitroanilide from D-Val-Leu-Lys-p-Nitroanilide per minute at pH 7.5 at 37 °C.

Physical form

Lyophilized powder containing sodium phosphate, mannitol, and NaCl.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Müllertz et al.
The Biochemical journal, 159(3), 545-553 (1976-12-01)
A complex between plasmin and an inhibitor was isolated by affinity chromatography from urokinase-activated human plasma. The complex did not react with antibodies against any of the known proteinase inhibitors in plasma. A rabbit antiserum against the complex was produced.
Efrat Shavit-Stein et al.
PloS one, 16(3), e0248431-e0248431 (2021-03-16)
Ischemic stroke is a common and debilitating disease with limited treatment options. Protease activated receptor 1 (PAR1) is a fundamental cell signaling mediator in the central nervous system (CNS). It can be activated by many proteases including thrombin and plasmin
Tatiana Syrovets et al.
Journal of leukocyte biology, 92(3), 509-519 (2012-05-09)
The serine protease plasmin generated from its zymogen plasminogen is best known for its function as a key enzyme of the fibrinolytic cascade. However, beyond fibrinolysis, plasmin has a number of crucial functions in a variety of processes, including inflammation.
Peter Stalmans et al.
Investigative ophthalmology & visual science, 54(10), 6620-6627 (2013-08-29)
To characterize the levels of active ocriplasmin over a period of ascending time points (range, 5 minutes to 7 days) from intravitreal injection of a 125-μg dose to sampling. During this 7-week controlled, open-label, phase 2 study, a single intravitreal
On the mechanism of the reaction between human alpha 2-antiplasmin and plasmin.
B Wiman et al.
The Journal of biological chemistry, 254(18), 9291-9297 (1979-09-25)

Protocols

Enzymatic Assay of Plasmin with D-Val-Leu-Lys-p-Nitroanilide Dihydrochloride

Protocol for Enzymatic Assay of Plasmin with D-Val-Leu-Lys-p-Nitroanilide Dihydrochloride

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service