Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C14H10N3Na4O12PS2 · xH2O
CAS Number:
Molecular Weight:
599.31 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Quality Segment
assay
≥98% (HPLC)
form
solid
color
red to orange
solubility
H2O: >10 mg/mL
storage temp.
−20°C
SMILES string
O.[Na+].[Na+].[Na+].[Na+].Cc1nc(\N=N\c2ccc(cc2S([O-])(=O)=O)S([O-])(=O)=O)c(COP([O-])([O-])=O)c(C=O)c1O
InChI
1S/C14H14N3O12PS2.4Na.H2O/c1-7-13(19)9(5-18)10(6-29-30(20,21)22)14(15-7)17-16-11-3-2-8(31(23,24)25)4-12(11)32(26,27)28;;;;;/h2-5,19H,6H2,1H3,(H2,20,21,22)(H,23,24,25)(H,26,27,28);;;;;1H2/q;4*+1;/p-4/b17-16+;;;;;
InChI key
JTTDHFFNVGHDLN-YDTRVRFHSA-J
Gene Information
Biochem/physiol Actions
Non-selective P2 purinoceptor antagonist which blocks responses at both pre- and post-junctional sites.
Features and Benefits
This compound is featured on the P2 Receptors: P2X Ion Channel Family and P2 Receptors: P2Y G-Protein Family pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
Disclaimer
Light sensitive
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.