Skip to Content

P1691

Poly(D,L-lactide)

mol wt 75,000-120,000

Synonym(s):

Poly[oxy(1-methyl-2-oxo-1,2-ethanediyl)]

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C3H4O2)n
CAS Number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

pellets

Quality Segment

mol wt

75,000-120,000

color

white to yellow

mp

262 °C (lit.)

transition temp

Tg 32.9 °C

storage temp.

−20°C

SMILES string

[O-]C(=O)C(O)C.[H+]

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Biochem/physiol Actions

Poly(D,L-lactide) is a polyamino acid that has been used to as a coating material.


Still not finding the right product?

Explore all of our products under Poly(D,L-lactide)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library