Skip to Content
Merck
All Photos(1)

Key Documents

P1584

Sigma-Aldrich

Peptidyl Arginine Deiminase from rabbit skeletal muscle

buffered aqueous glycerol solution, ≥200 units/mg protein (Bradford)

Synonym(s):

Protein arginine iminohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

buffered aqueous glycerol solution

Quality Level

specific activity

≥200 units/mg protein (Bradford)

relevant disease(s)

arthritis (rheumatoid )

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Peptidyl arginine deiminase is the enzyme that converts arginine into citrulline. [1]

Application

Peptidyl arginine deiminase has been used in a study that assessed promising novel biomarkers for the early diagnosis of rheumatoid arthritis. [2] It has also been used in a study to investigate the autopathogenic correlation of periodontitis and rheumatoid arthritis. [3]

Biochem/physiol Actions

Calcium is required for peptidylarginine deiminase activity in vitro. [4]

Unit Definition

One unit will produce 1 μmole of N-α-benzoylcitrulline ethyl ester from BAEE per hr at 55 °C at pH 7.2.

Physical form

Solution in 20 mM Tris-HCl, pH 7.4, containing 10 mM 2-mercaptoethanol, 1 mM EDTA and 10% glycerol

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

John G Routsias et al.
Rheumatology (Oxford, England), 50(7), 1189-1193 (2011-02-24)
Recently, a number of studies have pointed to a potential relationship between periodontitis (PO) and RA and vice versa. Both diseases are characterized by chronic inflammation, osseous destruction, damage of the supporting soft tissues, similar cellular immune responses and common
Murat Bozdag et al.
Bioorganic & medicinal chemistry letters, 23(3), 715-719 (2012-12-26)
Protein arginin deaminase 4 (PAD4) is a calcium dependent enzyme which catalyses the conversion of peptidyl-arginine into peptidyl-citrulline and is implicated in several diseases such as rheumatoid arthritis (RA) and cancer. Herein we report the discovery of novel small-molecule, non
M Fujisaki et al.
Journal of biochemistry, 89(1), 257-263 (1981-01-01)
An enzyme which catalyzes the coversion of arginyl residues to citrullyl residues in protein was obtained from the extract of the epidermis of newborn rats. The enzyme required Ca2+ for its activity. The enzyme activity was enhanced in the presence
Jason S Knight et al.
Circulation research, 114(6), 947-956 (2014-01-16)
Neutrophil extracellular trap (NET) formation promotes vascular damage, thrombosis, and activation of interferon-α-producing plasmacytoid dendritic cells in diseased arteries. Peptidylarginine deiminase inhibition is a strategy that can decrease in vivo NET formation. To test whether peptidylarginine deiminase inhibition, a novel
Maria A Christophorou et al.
Nature, 507(7490), 104-108 (2014-01-28)
Citrullination is the post-translational conversion of an arginine residue within a protein to the non-coded amino acid citrulline. This modification leads to the loss of a positive charge and reduction in hydrogen-bonding ability. It is carried out by a small

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service