Skip to Content
Merck
All Photos(2)

Key Documents

P1566

Sigma-Aldrich

Poly(L-lactide)

mol wt 85,000-160,000

Synonym(s):

Lactel® B6002-2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

form

pellets

mol wt

85,000-160,000

color

faint beige to beige

application(s)

cell analysis

storage temp.

−20°C

SMILES string

C[C@@H](O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m1/s1

InChI key

JVTAAEKCZFNVCJ-UWTATZPHSA-N

Biochem/physiol Actions

Poly-D-lysine hydrobromide is a synthetic amino acid that enhances cell adhesion to solid substrates. It has additionally been shown to eliminate prion proteins from infected cells.

Legal Information

Lactel is a registered trademark of Evonik Corp

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanchun Liu et al.
Tissue engineering. Part A, 25(11-12), 878-886 (2018-10-05)
This study compared side-by-side the impact of donor age on the production of tissue-engineered small intestine (TESI). Each age represents a specific period of life: E18 for fetuses, 5-day-old pups for neonates, 21-day-old rats for weanlings, and 6-week-old rats for

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service