Skip to Content
Merck
All Photos(3)

Documents

P1374

Sigma-Aldrich

Anti-Pro-BDNF antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-pro-Brain-derived Neurotrophic Factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, human, rat

technique(s)

western blot: 1:200 using rat glioma C6 and human neuroblastoma SH-SY5Y cell lines
western blot: 1:400 using recombinant mouse proBDNF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)
mouse ... Bdnf(12064)
rat ... Bdnf(24225)

Related Categories

General description

Does not cross react with pro-NGF, pro-NT-3, or mature BDNF.
The brain derived neurotrophic factor (BDNF) gene is mapped to human chromosome 11p14.1. The gene encodes a precursor protein, proBDNF. Mature BDNF (mBDNF) is synthesized by post-translational cleavage of proBDNF. Does not cross react with pro-NGF, pro-NT-3, or mature BDNF.

Immunogen

peptide corresponding to amino acid residues 72-88 of human BDNF (precursor). This sequence has 16/17 residues identical in mouse and rat.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Brain derived neurotrophic factor (BDNF) belongs to a family of secreted proteins called neurotrophins. Neurotrophins are synthesized as preproneurotrophin precursors and further processed to proneurotrophins and subsequently to neurotrophins. BDNF is expressed in the developing and the mature brain. The influence of BDNF is mediated by its binding to TrkB receptor and sometimes the receptor p75NTR. The downstream pathways promoted by BDNF include PI3K, MAPK and JAK/STAT. The most important role of BDNF is the regulation of synaptic transmission and plasticity, protein synthesis and phosphorylation of local proteins.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, with 1% bovine serum albumin and ≤0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Qiao et al.
Brain research, 1663, 29-37 (2017-03-13)
Major depressive disorder (MDD) is one of the most common psychiatric disorder, but the underlying mechanisms are largely unknown. Increasing evidence shows that brain-derived neurotrophic factor (BDNF) plays an important role in the structural plasticity induced by depression. Considering the
Guiping Xu et al.
Translational neuroscience, 13(1), 430-439 (2022-12-24)
As an intravenous anesthetic, propofol has been indicated to induce neurotoxicity in both animal and human brains. It is of great significance to better understand the potential mechanism of propofol-induced neurotoxicity to eliminate the side effects of propofol. Esketamine is
11p14. 1 microdeletions associated with ADHD, autism, developmental delay, and obesity.
Shinawi M, et al.
American Journal of Medical Genetics. Part A, 155(6), 1272-1280 (2011)
Li Duan et al.
PloS one, 8(9), e73768-e73768 (2013-09-17)
While aberrant activation of microglial cells was evidently involved in neuroinflammation and neurotoxicity in the neurodegenerative diseases such as Alzheimer's and Parkinson's disease, objective of study was to address if activated microglias deliver their effect by releasing pro-neurotrophins. By in
Electroacupuncture at the Baihui acupoint alleviates cognitive impairment and exerts neuroprotective effects by modulating the expression and processing of brain-derived neurotrophic factor in APP/PS1 transgenic mice.
Lin R
Molecular Medicine Reports, 13(2), 1611-1617 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service