Skip to Content
Merck

Skip To

P0610

Dounce tissue grinder pestle

Large clearance, working volume 7 mL

Synonym(s):

885301-0007

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
eCl@ss:
32080115
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

glass

working volume

7 mL

General description

Designed primarily for cellular work where the nucleus remains intact after homogenization.

Features and Benefits

  • Manufactured from borosilicate glass 3.3
  • Designed primarily for cellular work where the nucleus remains intact after homogenization.
  • All-glass construction
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completely interchangeable


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P0485P1610P0735
material

glass

material

glass

material

glass

material

glass

working volume

7 mL

working volume

2 mL

working volume

100 mL

working volume

15 mL


Still not finding the right product?

Explore all of our products under Dounce tissue grinder pestle




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library