Skip to Content
Merck
All Photos(4)

Key Documents

P0547

Sigma-Aldrich

Pentamidine isethionate salt

powder

Synonym(s):

1,5-Bis(p-amidinophenoxy)pentane bis(2-hydroxyethanesulfonate salt), 4,4′-(Pentamethylenedioxy)dibenzamidine bis(2-hydroxyethanesulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[CH2CH2OC6H4C(=NH)NH2]2·2HOCH2CH2SO3H
CAS Number:
Molecular Weight:
592.68
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

mp

188-194 °C (lit.)

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

originator

Celgene

storage temp.

−20°C

SMILES string

OCCS(O)(=O)=O.OCCS(O)(=O)=O.NC(=N)c1ccc(OCCCCCOc2ccc(cc2)C(N)=N)cc1

InChI

1S/C19H24N4O2.2C2H6O4S/c20-18(21)14-4-8-16(9-5-14)24-12-2-1-3-13-25-17-10-6-15(7-11-17)19(22)23;2*3-1-2-7(4,5)6/h4-11H,1-3,12-13H2,(H3,20,21)(H3,22,23);2*3H,1-2H2,(H,4,5,6)

InChI key

YBVNFKZSMZGRAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Antimicrobial against Pneumocystis carinii.
Pentamidine isethionate salt has been used:
  • as a positive control for anti-leishmanial, anti-plasmodial and cytotoxic assays
  • as a prototypical compound to analyse the inhibitor effect on human ether-a-go-go-related gene (HERG) K+ channel trafficking
  • as an anti-leishmanial agent in promastigotes

Biochem/physiol Actions

Neuroprotective; inhibits constitutive nitric oxide synthase in the brain; NMDA glutamate receptor antagonist.
Pentamidine isethionate is used as a second line drug for leishmaniasis.

Features and Benefits

This compound was developed by Celgene. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I J Reynolds et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 12(3), 970-975 (1992-03-01)
Acquired immunodeficiency syndrome (AIDS) is frequently associated with dementia. The wide spectrum of neurological abnormalities associated with this dementia may involve a neurotoxin that activates the NMDA subtype of glutamate receptor in neurons. We have found that the antimicrobial agent
Anti-leishmanial lindenane sesquiterpenes from Hedyosmum angustifolium
Acebey L, et al.
Planta Medica, 76(04), 365-368 (2010)
Drug-induced functional cardiotoxicity screening in stem cell-derived human and mouse cardiomyocytes: effects of reference compounds
Himmel HM
Journal of Pharmacological and Toxicological Methods, 68(1), 97-111 (2013)
Reduced Antimony Accumulation in ARM58-Overexpressing Leishmania infantum.
Schafer C, et al.
Antimicrobial Agents and Chemotherapy, 58(3), 1565-1574 (2014)
Lucia Acebey et al.
Planta medica, 76(4), 365-368 (2009-10-16)
The aim of this work is the isolation of anti-leishmanial compounds from the ethyl acetate extracts of the bark of HEDYOSMUM ANGUSTIFOLIUM. We have successfully isolated and characterized five sesquiterpenes: one new compound (oxyonoseriolide, 1), one compound isolated for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service