Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
assay
≥97% (HPLC)
form
solid
storage condition
desiccated
solubility
DMSO: soluble >10 mg/mL, clear, H2O: insoluble
storage temp.
2-8°C
SMILES string
NS(=O)(=O)C#Cc1ccccc1
InChI
1S/C8H7NO2S/c9-12(10,11)7-6-8-4-2-1-3-5-8/h1-5H,(H2,9,10,11)
InChI key
ZZUZYEMRHCMVTB-UHFFFAOYSA-N
Application
- to treat microglial cell line to analyse its neuroprotective effect on M1-like and M2-like phenotype
- as heat shock protein (HSP)-70 inhibitor, to treat transfected Marc-145 cells
- to inhibit heat shock cognate 70 (Hsc70) to elucidate heat shock chaperones mouse embryonic stem cells
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form solid | form solid | form solid |
| assay ≥97% (HPLC) | assay ≥95% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| storage condition desiccated | storage condition OK to freeze, protect from light | storage condition - | storage condition - |
| solubility DMSO: soluble >10 mg/mL, clear, H2O: insoluble | solubility DMSO: 50 mg/mL | solubility DMSO: >10 mg/mL, H2O: insoluble | solubility DMSO: >10 mg/mL, H2O: insoluble |
Still not finding the right product?
Explore all of our products under Pifithrin-μ
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




